  • Search Results
  • Development of a subunit vaccine containing recombinant Riemerella anatipestifer outer membrane protein A and CpG ODN adjuvant.
Development of a subunit vaccine containing recombinant Riemerella anatipestifer outer membrane protein A and CpG ODN adjuvant.
Vaccine (2014-12-03)
Chun-Yen Chu, Chia-Hui Liu, Jhong-Jie Liou, Jai-Wei Lee, Li-Ting Cheng
ABSTRACT
Riemerella anatipestifer, a Gram-negative bacillus, causes septicemia that can result in high mortality for ducklings. In this study, we evaluated the immune response and protective efficacy provided by a subunit vaccine containing recombinant outer membrane protein A (rOmpA) and plasmid constructs containing CpG oligodeoxynucleotides (ODN). Results showed that CpG ODN enhanced both humoral and cell-mediated immunity elicited by rOmpA as early as two weeks after primary immunization. When compared to ducks immunized with rOmpA, ducks immunized with rOmpA+CpG ODN showed higher levels (p<0.05) of antibody titer, T cell proliferation, and percentages of CD4(+) and CD8(+) T cell in peripheral blood mononuclear cells (PBMCs). The relative fold inductions of mRNA expression of Th1-type (IFN-γ and IL-12), and Th2-type (IL-6) cytokines in PBMCs isolated from ducks immunized with rOmpA+CpG ODN were significantly higher than those of the rOmpA group. Homologous challenge result showed that the rOmpA+CpG ODN vaccine reduced the pathological score by 90% in comparison with the saline control. In conclusion, our study found that CpG ODN can enhance both humoral and cellular immunity elicited by a rOmpA vaccine. The rOmpA+CpG ODN vaccine can be further developed as a subunit vaccine against R. anatipestifer.
Sigma-Aldrich
IPTG, ≥99% (TLC), ≤0.1% Dioxane
Sigma-Aldrich
Isopropyl β-D-1-thiogalactopyranoside, ≥99% (TLC)
SAFC
Isopropyl β-D-1-thiogalactopyranoside
Sigma-Aldrich
Isopropyl β-D-thiogalactopyranoside solution, ReadyMade IPTG solution for Blue-white screening
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
Sodium azide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium azide, BioXtra