  • Search Results
  • Human megakaryocyte progenitors derived from hematopoietic stem cells of normal individuals are MHC class II-expressing professional APC that enhance Th17 and Th1/Th17 responses.
Human megakaryocyte progenitors derived from hematopoietic stem cells of normal individuals are MHC class II-expressing professional APC that enhance Th17 and Th1/Th17 responses.
Immunology letters (2014-12-03)
Ariel Finkielsztein, Alaina C Schlinker, Li Zhang, William M Miller, Syamal K Datta
ABSTRACT
Platelets, like stromal cells, present antigen only via MHC class I, but the immune potential of their progenitors has not been explored in humans. We derived CD34(+)CD117(+)CD41(+)CD151(+) megakaryocyte progenitors (MKp) in vitro from mobilized peripheral blood hematopoietic stem and progenitor cells (HSPC) of normal subjects using culture conditions akin to bone marrow niche, or organs that support extramedullary hematopoiesis. The MKp expressed MHC Class II in contrast to platelets and functioned as professional APC before they matured further. Moreover, MKp constitutively expressed mRNA encoding mediators for human Th17 expansion, including IL-1, IL-18, IL-6, TGFβ, IL-23, BAFF, and COX2. MKp also expressed high levels of type I interferon and IRF5 mRNA. In contrast to platelets, MKp augmented the expansion of Th17, Th1, and potent Th17/Th1 double-positive cells in normal PBMC and CD4 line T cells from normal subjects or lupus patients. The Th cell augmentation involved pre-committed memory cells, and was significant although modest, because only non-cognate MKp-T cell interactions could be studied, under non-polarizing conditions. Importantly, the MKp-mediated expansion was observed in the presence or absence of direct MKp-T cell contact. Furthermore, MKp augmented Th17 responses against Candida albicans, a serious opportunistic pathogen. These results indicate an immunologic role of MKp in situations associated with extramedullary hematopoiesis and mobilization of HSPC.
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, synthetic, ≥98.0% (TLC)
Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
Aphidicolin from Nigrospora sphaerica, ≥98% (HPLC), powder
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, ≥99% (TLC), film or powder
Supelco
Aphidicolin, analytical standard
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
SAFC
HEPES
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
SAFC
HEPES
Supelco
HEPES, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethanol, purum, secunda spirit, denaturated with 2% 2-butanone and 0.5% 4-methyl-2-pentanone, S15, ~96% (based on denaturant-free substance)
Supelco
Ethanol, standard for GC
Sigma-Aldrich
Ethanol, BioUltra, for molecular biology, ≥99.8%, (absolute alcohol, without additive, A15 o1)
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS reagent, meets USP testing specifications, Excise Tax-free, Permit for use required
Sigma-Aldrich
Ethyl alcohol, Pure, 160 proof, Excise Tax-free, Permit for use required
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, for molecular biology
Sigma-Aldrich
Ethanol, purum, absolute ethanol, denaturated with 4.8% isopropanol, A15 IPA1, ≥99.8% (based on denaturant-free substance)
Sigma-Aldrich
Ethanol, ACS reagent, prima fine spirit, without additive, F15 o1
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 2% 2-butanone, F25 MEK1, ~96% (based on denaturant-free substance)
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, HPLC/spectrophotometric grade