  • Search Results
  • Insights into the inhibitory mechanisms of the regulatory protein IIA(Glc) on melibiose permease activity.
Insights into the inhibitory mechanisms of the regulatory protein IIA(Glc) on melibiose permease activity.
The Journal of biological chemistry (2014-10-10)
Parameswaran Hariharan, Lan Guan
ABSTRACT
The phosphotransfer protein IIA(Glc) of the bacterial phosphoenolpyruvate:carbohydrate phosphotransferase system plays a key role in the regulation of carbohydrate metabolism. Melibiose permease (MelB) is one among several permeases subject to IIA(Glc) regulation. The regulatory mechanisms are poorly understood; in addition, thermodynamic features of IIA(Glc) binding to other proteins are also unknown. Applying isothermal titration calorimetry and amine-specific cross-linking, we show that IIA(Glc) directly binds to MelB of Salmonella typhimurium (MelB(St)) and Escherichia coli MelB (MelB(Ec)) at a stoichiometry of unity in the absence or presence of melibiose. The dissociation constant values are 3-10 μM for MelB(St) and 25 μM for MelB(Ec). All of the binding is solely driven by favorable enthalpy forces. IIA(Glc) binding to MelB(St) in the absence or presence of melibiose yields a large negative heat capacity change; in addition, the conformational entropy is constrained upon the binding. We further found that the IIA(Glc)-bound MelB(St) exhibits a decreased binding affinity for melibiose or nitrophenyl-α-galactoside. It is believed that sugar binding to the permease is involved in an induced fit mechanism, and the transport process requires conformational cycling between different states. Thus, the thermodynamic data are consistent with the interpretation that IIA(Glc) inhibits the induced fit process and restricts the conformational dynamics of MelB(St).
Product Description
Ondansetron impurity E, European Pharmacopoeia (EP) Reference Standard
Imidazole, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
Imidazole, ACS reagent, ≥99% (titration)
Sigma-Aldrich
Imidazole, ≥99% (titration), crystalline
Sigma-Aldrich
Imidazole, for molecular biology, ≥99% (titration)
Sigma-Aldrich
Imidazole, ReagentPlus®, 99%
Sigma-Aldrich
Imidazole, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Imidazole, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Imidazole, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Imidazole, for molecular biology, ≥99% (titration), free-flowing, Redi-Dri
Sigma-Aldrich
Imidazole, ReagentPlus®, 99%, Redi-Dri, free-flowing
Supelco
Imidazole, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Imidazole, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Sigma-Aldrich
Sucrose, Grade II, suitable for plant cell culture
Sigma-Aldrich
Sucrose, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Imidazole buffer Solution, BioUltra, 1 M in H2O
Supelco
Sucrose, analytical standard
Millipore
Sucrose, ACS reagent, suitable for microbiology, ≥99.0%
Sigma-Aldrich
Sucrose, BioUltra, for molecular biology, ≥99.5% (HPLC)
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Supelco
Sucrose, Pharmaceutical Secondary Standard; Certified Reference Material
Sucrose, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sucrose, ACS reagent
Supelco
Sucrose, analytical standard, for enzymatic assay kit SCA20
Sigma-Aldrich
Sucrose, meets USP testing specifications
Sigma-Aldrich
Sucrose, ≥99.5%
Sigma-Aldrich
Sucrose, for molecular biology, ≥99.5% (GC)