• Início
  • Resultados da busca
  • Ultra-high performance liquid chromatography-tandem mass spectrometry for the quantification of icaritin in mouse bone.
Ultra-high performance liquid chromatography-tandem mass spectrometry for the quantification of icaritin in mouse bone.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2014-12-23)
Shuang-Qing Zhang
RESUMO
Icaritin (ICT), a bioactive metabolite of prenylflavonoids from genus Epimedium, has displayed potential benefits for the treatment of osteoporosis, prostate cancer, liver cancer, renal cancer and breast cancer. To investigate the quantity of ICT in bones in vivo, a simple and sensitive ultra-high performance liquid chromatography-tandem mass spectrometry (UHPLC-MS/MS) method was developed. After a rapid one-step liquid-liquid extraction using ethyl acetate with recovery more than 87.2% at four levels (0.1, 0.2, 8 and 15 ng/mL), ICT and internal standard coumestrol were analyzed on a C18 column using a gradient elution of acetonitrile and water containing ammonium formate and formic acid at a flow rate of 0.3 mL/min. Quantification was performed using selected reaction monitoring mode to monitor precursor-product ion transitions of m/z 367.1→297.1 for ICT and of 267.0→211.1 for coumestrol in the negative ionization mode. A calibration curve with good linearity (r>0.99) within the concentration range of 0.1-20 ng/mL for ICT was obtained with the lower limit of quantification of 0.1 ng/mL. Matrix effect did not interfere with ICT analysis and ICT was stable under three freeze-thaw cycles, short-term temperature, post-preparative and long-term temperature conditions. The method was successfully applied to a dynamic distribution of ICT in mouse bone after a single intraperitoneal administration to ICT to mice.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Ethyl acetate, ACS reagent, ≥99.5%
Supelco
Ammonium formate, eluent additive for LC-MS, LiChropur, ≥99.0%
Sigma-Aldrich
Ethyl acetate, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Ethyl acetate, anhydrous, 99.8%
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Formic acid, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥98%
Sigma-Aldrich
Acetonitrile, for HPLC, for UV, ≥99.9% (GC)
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Ammonium formate, reagent grade, 97%
Sigma-Aldrich
Ethyl acetate, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
Ammonium formate, ≥99.995% trace metals basis
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Formic acid, puriss., meets analytical specifications of DAC, FCC, 98.0-100%
Sigma-Aldrich
Ammonium formate, BioUltra, ≥99.0% (calc. based on dry substance, NT)
Sigma-Aldrich
Ethyl acetate, puriss., meets analytical specification of Ph. Eur., BP, NF, ≥99.5% (GC)
Sigma-Aldrich
Ammonium formate solution, BioUltra, 10 M in H2O
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Supelco
Ethyl acetate, analytical standard
Sigma-Aldrich
Ethyl acetate, suitable for HPLC, ≥99.8%
Sigma-Aldrich
Ethyl acetate, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile, suitable for HPLC-GC, ≥99.8% (GC)
Supelco
Ethyl Acetate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Supelco
Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material