Merck
Simple and reliable HPLC method for the monitoring of methotrexate in osteosarcoma patients.
Journal of chromatographic science (2013-06-27)
Elias Begas, Christos Papandreou, Andreas Tsakalof, Danai Daliani, George Papatsibas, Eftihia Asprodini
RESUMO
Methotrexate (MTX) is a dihydrofolate reductase inhibitor that is used for the treatment of tumors and autoimmune diseases. Several automated binding assays are used in clinical practice and numerous chromatographic methods have been developed toward higher specificity and sensitivity. In the present study, phenyl cartridges were used for the solid-phase extraction (SPE) of MTX from human serum samples; subsequently, extracts were analyzed by reversed-phase high-performance liquid chromatography. Isocratic separation was implemented on a Kromasil-C18 column with a mobile phase consisting of 50 mM sodium acetate buffer (pH 3.6)-acetonitrile (89:11, v/v) and ultraviolet detection at 307 nm. MTX eluted in less than 12 min with no interference from impurities or 24 examined drugs. Detector response was linear in the range of 0.025-5.00 µΜ (coefficient of correlation > 0.99). Recovery from the serum was 93.1-98.2% and bias was < 8.3%. Intra-day and inter-day precision were <7.8 and 12.6%, respectively (n = 6). The limit of quantitation was 0.01 µM and the limit of detection was 0.003 µΜ. The method was validated by using serum samples from osteosarcoma patients treated with high-dose MTX (8-12 g/m(2)). In conclusion, the combined use of a phenyl-functionalized sorbent for SPE and a Kromasil-C18 column, and specific detection at 307 nm, assured a selective, fast, robust and cost-effective method for the monitoring of MTX in osteosarcoma patients under high-dose MTX treatment, thus contributing to more efficient treatment.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Sodium Acetate Anhydrous, >99%, FCC, FG
Supelco
Sodium acetate solution, NMR reference standard, 50 mM in D2O (99.9 atom % D), water 1 %, NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Sodium acetate, anhydrous, BioUltra, for luminescence, for molecular biology, ≥99.0% (NT)
Supelco
Acetic acid, analytical standard
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Sodium acetate, 99.995% trace metals basis
Sigma-Aldrich
Sodium acetate solution, BioUltra, for molecular biology, ~3 M in H2O
Sigma-Aldrich
Sodium acetate, meets USP testing specifications, anhydrous
Sigma-Aldrich
Sodium acetate, anhydrous, for molecular biology, ≥99%
Sigma-Aldrich
Sodium acetate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium acetate, powder, BioReagent, suitable for electrophoresis, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Paracetamol, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Sodium acetate, anhydrous, ReagentPlus®, ≥99.0%
Methotrexate for system suitability, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Acetic acid, glacial, puriss., 99-100%
Sigma-Aldrich
Acetic acid, glacial, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, 99.8-100.5%
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Acetic acid, glacial, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8%
Sigma-Aldrich
Sodium acetate, puriss. p.a., ACS reagent, reag. Ph. Eur., anhydrous
Sigma-Aldrich
Sodium acetate, ACS reagent, ≥99.0%
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Sigma-Aldrich
Sodium acetate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.0%
Sigma-Aldrich
Methotrexate, meets USP testing specifications
Sigma-Aldrich
Acetaminophen, BioXtra, ≥99.0%
Sigma-Aldrich
Acetaminophen, meets USP testing specifications, 98.0-102.0%, powder