• Início
  • Resultados da busca
  • Plant regeneration and biochemical accumulation of hydroxybenzoic and hydroxycinnamic acid derivatives in Hypoxis hemerocallidea organ and callus cultures.
Plant regeneration and biochemical accumulation of hydroxybenzoic and hydroxycinnamic acid derivatives in Hypoxis hemerocallidea organ and callus cultures.
Plant science : an international journal of experimental plant biology (2014-09-16)
Mack Moyo, Stephen O Amoo, Adeyemi O Aremu, Jiří Gruz, Michaela Subrtová, Karel Doležal, Johannes Van Staden
RESUMO
Micropropagation of Hypoxis hemerocallidea Fisch. and C.A. Mey was used as a model system to study the influence of cytokinins (CKs) on plant regeneration and biochemical accumulation of hydroxybenzoic and hydroxycinnamic acid derivatives in organ and callus cultures and their antioxidant activity. Fourteen free phenolic acids were detected using ultra performance liquid chromatography-tandem mass spectrometry (UPLC-MS/MS) while antioxidant activity was evaluated using oxygen radical absorbance capacity (ORAC) and 2,2-diphenyl-1-picryl hydrazyl (DPPH) radical scavenging activity. Cytokinins had a significant effect on the biochemical accumulation of hydroxybenzoic and hydroxycinnamic acid derivatives in H. hemerocallidea organ cultures. In particular, meta-topolin-treated organ cultures produced high concentrations of gallic, protocatechuic, gentisic, p-hydroxybenzoic, m-hydroxybenzoic, salicylic, chlorogenic and trans-cinnamic acids. The isoprenoid CK, N(6)-(2-isopentenyl)-adenine significantly increased the accumulation of hydroxycinnamic acid derivatives, namely, caffeic, p-coumaric, sinapic and ferulic acids. Cytokinin-treated organ cultures exhibited a significant increase in antioxidant activity, particularly in the ORAC model. In callus cultures, CKs decreased the concentrations of hydroxycinnamic acid derivatives and antioxidant activity when compared to the control. Overall, both CK type and concentration had a significant effect on plant regeneration, callus proliferation, biochemical accumulation of free phenolic acids and antioxidant activity of the resultant extracts.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Supelco
Methanol, analytical standard
Sigma-Aldrich
Glycine, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Formic acid, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥98%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
(±)-α-Tocopherol, synthetic, ≥96% (HPLC)
Sigma-Aldrich
Fluorescein sodium salt, used as fluorescent tracer
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
myo-Inositol, ≥99%
Sigma-Aldrich
Glycine, BioUltra, for molecular biology, ≥99.0% (NT)
Supelco
Alpha Tocopherol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
myo-Inositol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Nicotinic acid, ≥98%
Sigma-Aldrich
Glycine, ACS reagent, ≥98.5%
Sigma-Aldrich
Formic acid, puriss., meets analytical specifications of DAC, FCC, 98.0-100%
Sigma-Aldrich
Glycine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Sigma-Aldrich
α-Tocopherol, ≥95.5%
Supelco
(±)-α-Tocopherol, analytical standard
Sigma-Aldrich
Fluorescein sodium salt, BioReagent, suitable for fluorescence
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Pyridoxine, ≥98%
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%