Multi-residues determination of antimicrobials in fish tissues by HPLC-ESI-MS/MS method.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2014-12-23)
Mamdouh R Rezk, Safa'a M Riad, Fatma I Khattab, Hoda M Marzouk
RESUMO
A rapid, simple, sensitive and specific LC-MS/MS method was developed and validated for the simultaneous quantification of four antimicrobials commonly used in aquaculture, namely ciprofloxacin (CPX), trimethoprim (TMP), sulphadimethoxine (SDM) and florphenicol (FLOR) in fish tissues. The LC-MS/MS was operated under the multiple-reaction monitoring mode using electrospray ionization. Sample preparation involves simple liquid extraction step followed by post-extraction clean-up step with n-hexane. The purified extracts were chromatographed on Agilent Poroshell 120 EC, C18 (50 mm × 3 mm, 2.7 μm) column by pumping an isocratic mobile phase consisting of 0.1% formic acid in water:0.1% formic acid in methanol (20:80, by volume) at a flow rate of 0.4 mL/min. A detailed validation of the method was performed as per FDA guidelines and the standard curves were found to be linear in the range of 1-100 ng/g for both CPX and TMP, 0.5-100 ng/g for SDM and 1-50 ng/g for FLOR. The intra-day and inter-day precision and accuracy of the results were within the acceptable limits. A run time of 1.5 min for each sample made it possible to analyze multiple fish tissue samples per day. The developed assay method was successfully applied for the detection of antimicrobials in real fish tissue samples obtained from different fish farms.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Supelco
Methanol, analytical standard
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Formic acid, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥98%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Acetonitrile, for HPLC, for UV, ≥99.9% (GC)
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Formic acid, puriss., meets analytical specifications of DAC, FCC, 98.0-100%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for HPLC-GC, ≥99.8% (GC)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Supelco
Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material