• Início
  • Resultados da busca
  • In vitro study of the interaction of heregulin-functionalized magnetic-optical nanorods with MCF7 and MDA-MB-231 cells.
In vitro study of the interaction of heregulin-functionalized magnetic-optical nanorods with MCF7 and MDA-MB-231 cells.
Faraday discussions (2014-10-01)
A Lesniak, D Kilinc, Suad A Rashdan, A von Kriegsheim, B Ashall, D Zerulla, W Kolch, G U Lee
RESUMO
Multifunctional nanoparticles that actively target specific cells are promising tools for cancer diagnosis and therapy. In this article we review the synthesis and surface chemistry of Fe-Au nanorods and their characterization using microscopy. The diameter of the rods used in this study was selected to be 150-200 nm so that they did not enter the cells. The 80 nm-long Au tips of the nanorods were functionalized with heregulin (HRG), and the micron-long Fe portion was coated with a poly(ethylene glycol) monolayer to minimize non-specific interactions. Nanorods functionalized with HRG were found to preferentially bind to MCF7 cells that express high levels of the receptor tyrosine-protein kinase ErbB2/3. Magnetic tweezers measurements were used to characterize the kinetic properties of the bond between the HRG on the rods and ErbB2/3 on the surface of the cells. The strong magnetization of Fe-Au nanorods makes them excellent candidates for in-vitro and in-vivo imaging, and magnetic therapeutic applications targeting cancer cells in circulation.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Triethylamine, ≥99.5%
Sigma-Aldrich
N-(3-Dimethylaminopropyl)-N′-ethylcarbodiimide hydrochloride, commercial grade, powder
Sigma-Aldrich
Triethylamine, ≥99%
Sigma-Aldrich
1,2-Dichloroethane, anhydrous, 99.8%
Sigma-Aldrich
N-(3-Dimethylaminopropyl)-N′-ethylcarbodiimide hydrochloride, purum, ≥98.0% (AT)
Sigma-Aldrich
N-Hydroxysulfosuccinimide sodium salt, ≥98% (HPLC)
Sigma-Aldrich
Triethylamine, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
N-(3-Dimethylaminopropyl)-N′-ethylcarbodiimide hydrochloride, crystalline
Sigma-Aldrich
1,2-Dichloroethane, ACS reagent, ≥99.0%
Sigma-Aldrich
Triethylamine, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
N-(3-Dimethylaminopropyl)-N′-ethylcarbodiimide hydrochloride, BioXtra
Sigma-Aldrich
N-(3-Dimethylaminopropyl)-N′-ethylcarbodiimide hydrochloride, ≥99.0% (AT)
Sigma-Aldrich
1,2-Dichloroethane, suitable for HPLC, ≥99.8%
Sigma-Aldrich
MES solution, BioUltra, for molecular biology, 0.5 M in H2O
Supelco
1,2-Dichloroethane, analytical standard
Supelco
Triethylamine, analytical standard
Sigma-Aldrich
Triethylamine, for amino acid analysis, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, for protein sequence analysis, ampule, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, ≥99.5%