  • Search Results
  • Expression and epigenetic regulation of DACT1 and DACT2 in oral squamous cell carcinoma.
Expression and epigenetic regulation of DACT1 and DACT2 in oral squamous cell carcinoma.
Cancer biomarkers : section A of Disease markers (2014-12-20)
Juliana Lucena Schussel, Luciana Puchalski Kalinke, Laurindo Moacir Sassi, Benedito Valdecir de Oliveira, Paola A G Pedruzzi, Marcia Olandoski, Lucia Elvira Alvares, Gustavo Pompermaier Garlet, Paula Cristina Trevilatto
ABSTRACT
DACT genes regulates Wnt as well TGF-β pathway, and were already associated with hepatocellular and lung cancer. Alterations on Wnt/β-catenin were associated with head and neck cancer through β-catenin cytoplasmatic accumulation. The aim of the study was to evaluate DACT1 and DACT2 expression and methylation on oral squamous cell cancer (OSCC). 47 samples of salivary rinse and tissue were collected from 29 OSCC and 18 control patients. qMSP and RT-PCR reactions were performed in order to detect hypermethylation and expression of DACT1 and DACT2 genes. Statistical analysis was conducted to evaluate these genes as possible biomarkers for OSCC. As expected man over 60 years old with tobacco and alcohol consumption history were associated with OSCC. There was no statistical difference between groups concerning DACT1 and DACT2 either in promoter hypermethylation or transcript levels. Age was associated with DACT2 promoter hypermethylation, especially over 56 years old. Patients older than 56 years old were about 5 times more likely to have DACT2 promoter hypermethylation. These findings could partially explain why older subjects are more prone to carcinogenesis. Wnt/β-catenin pathway plays an important role in carcinogenesis, and the study of their regulators may help understand malignant transformation.
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Ammonium-14N2 sulfate solution, 40 wt. % in H2O, 99.99 atom % 14N
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90%
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Ammonium-14N2,sulfate-16O4, 99.99 atom % 14N, 99.99 atom % 16O
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Ammonium sulfate-14N2 solution, 40 wt. % in H2O, 99.99 atom % 14N
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Ammonium-14N2 sulfate, 99.99 atom % 14N
Sigma-Aldrich
Ammonium sulfate, for molecular biology, ≥99.0%
Sigma-Aldrich
Ammonium sulfate, 99.999% trace metals basis
Sigma-Aldrich
Ammonium sulfate, BioUltra, ≥99.0% (T)
Supelco
Ammonium sulfate, analytical standard, for Nitrogen Determination According to Kjeldahl Method, traceable to NIST SRM 194
Sigma-Aldrich
Ammonium sulfate, BioXtra, ≥99.0%
Sigma-Aldrich
Ammonium sulfate, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Ammonium sulfate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.0%
Sigma-Aldrich
Ammonium sulfate, anhydrous, Redi-Dri, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Ammonium sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Ammonium sulfate, ReagentPlus®, ≥99.0%
Sodium laurilsulfate, European Pharmacopoeia (EP) Reference Standard