  • Search Results
  • Evaluation of the antioxidant activity of Coreopsis tinctoria Nuff. and optimisation of isolation by response surface methodology.
Evaluation of the antioxidant activity of Coreopsis tinctoria Nuff. and optimisation of isolation by response surface methodology.
Acta pharmaceutica (Zagreb, Croatia) (2014-10-10)
Shumin Lan, Jingming Lin, Ni Zheng
ABSTRACT
The aim of the present study was to determine the antioxidant activity of Coreopsis tinctoria flowering tops (CTFT). Studies were conducted to obtain suitable extraction conditions for chlorogenic acid, quercetin, luteolin, apigenin and kaempferol, which were identified and quantified by HPLC. Response surface methodology was employed to optimise the ultrasound-assisted extraction conditions including extraction time, ethanol concentration and liquid-solid ratio. The antioxidant activity of the extracts was analysed using various antioxidant models, such as DPPH, ABTS and hydroxyl radical scavenging assay. CTFT extracted for 15.0 min with ethanol at a concentration of 60.4% and with liquidsolid ratio 27.5:1 possessed a considerable amounts of total flavonoids and polyphenols (18.9%). This extract showed higher scavenging activity of ABTS and hydroxyl radical activity than rutin, however not in the DPPH test. We may assume that CTFT possess antioxidant and free radical scavenging potentials.
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
L-Ascorbic acid, tested according to Ph. Eur.
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, suitable for plant cell culture
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Supelco
L-Ascorbic acid, certified reference material, TraceCERT®
Supelco
L-Ascorbic acid, analytical standard
Sigma-Aldrich
2,2-Diphenyl-1-picrylhydrazyl
Sigma-Aldrich
1,1-Diphenyl-2-picrylhydrazine, 97%
Ascorbic acid, European Pharmacopoeia (EP) Reference Standard
Supelco
Ascorbic Acid, Pharmaceutical Secondary Standard; Certified Reference Material