Cytochrome P450 OxyBtei catalyzes the first phenolic coupling step in teicoplanin biosynthesis.
Chembiochem : a European journal of chemical biology (2014-11-02)
Kristina Haslinger, Egle Maximowitsch, Clara Brieke, Alexa Koch, Max J Cryle
RESUMO
Bacterial cytochrome P450s form a remarkable clade of the P450 superfamily of oxidative hemoproteins, and are often involved in the biosynthesis of complex natural products. Those in a subgroup known as "Oxy enzymes" play a crucial role in the biosynthesis of glycopeptide antibiotics, including vancomycin and teicoplanin. The Oxy enzymes catalyze crosslinking of aromatic residues in the non-ribosomal antibiotic precursor peptide while it remains bound to the non-ribosomal peptide synthetase (NRPS); this crosslinking secures the three-dimensional structure of the glycopeptide, crucial for antibiotic activity. We have characterized OxyBtei , the first of the Oxy enzymes in teicoplanin biosynthesis. Our results reveal that OxyBtei possesses a structure similar to those of other Oxy proteins and is active in crosslinking NRPS-bound peptide substrates. However, OxyBtei displays a significantly altered activity spectrum against peptide substrates compared to its well-studied vancomycin homologue.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Formic acid, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥98%
Sigma-Aldrich
Acetonitrile, for HPLC, for UV, ≥99.9% (GC)
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Formic acid, puriss., meets analytical specifications of DAC, FCC, 98.0-100%
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile, suitable for HPLC-GC, ≥99.8% (GC)
Supelco
Ketoconazole, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Clotrimazole, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ketoconazole, 99.0-101.0% (EP, titration)
Sigma-Aldrich
Clotrimazole
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Supelco
Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
4-Chloro-DL-phenylalanine
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Sigma-Aldrich
Ketoconazole
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Supelco
Clotrimazole, VETRANAL®, analytical standard