  • Search Results
  • The contribution of human OCT1, OCT3, and CYP3A4 to nitidine chloride-induced hepatocellular toxicity.
The contribution of human OCT1, OCT3, and CYP3A4 to nitidine chloride-induced hepatocellular toxicity.
Drug metabolism and disposition: the biological fate of chemicals (2014-04-30)
Liping Li, Meijuan Tu, Xi Yang, Siyuan Sun, Xiaodan Wu, Hui Zhou, Su Zeng, Huidi Jiang
ABSTRACT
Nitidine chloride (NC), a quaternary ammonium alkaloid, has numerous pharmacological effects, such as anticancer activity. However, it was found that NC also has hepatocellular toxicity. Because organic cation transporters 1 and 3 (OCT1 and OCT3) might mediate the influx of NC into hepatocytes, multidrug and toxin extrusion 1 (MATE1) probably mediates the efflux of NC from hepatocytes, while cytochrome P450 (P450) enzymes might contribute to NC metabolism, the present study was to evaluate the contribution of OCT1, OCT3, MATE1, and P450 enzymes to NC-induced hepatocellular toxicity. Our results showed that the uptake of NC in Madin-Darby canine kidney (MDCK) cells expressing human (h) OCT1 and OCT3 (MDCK-hOCT1 and MDCK-hOCT3) was significantly higher than that in mock cells; the hOCT1- and hOCT3-mediated uptake followed typical Michaelis-Menten kinetics. Meanwhile, NC was also a substrate of hMATE1, although its transport capacity was much lower than that of OCT1 NC-induced cytotoxicity in MDCK-hOCT1 or MDCK-hOCT3 cells was obviously higher than that in mock cells. Quinidine and (+)-tetrahydropalmatine [(+)-THP], OCT1 and OCT3 inhibitors, significantly reduced the uptake of NC in MDCK-hOCT1 cells, MDCK-hOCT3 cells, and rat primary hepatocytes, but only (+)-THP markedly attenuated the NC-induced toxicity. In addition, P450 enzymes, such as CYP3A4, mediated the metabolism of NC, and NC-induced toxicity in MDCK-hOCT1/hCYP3A4 cells was lower than that in MDCK-hOCT1 cells. Our results indicated that NC is a substrate of hOCT1, hOCT3, and CYP3A4; that OCT1 and OCT3 mediate the uptake of NC in hepatocytes and subsequently cause hepatotoxicity; and that NC-induced toxicity could be attenuated by CYP3A4-mediated metabolism.
Sigma-Aldrich
Quinidine, anhydrous
Sigma-Aldrich
Furafylline, ≥98% (HPLC)
Sigma-Aldrich
Ketoconazole, 99.0-101.0% (EP, titration)
Sigma-Aldrich
Sulfaphenazole, ≥98%
Sigma-Aldrich
Calcium chloride, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Sigma-Aldrich
Calcium chloride, meets USP testing specifications
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.99% trace metals basis
Sigma-Aldrich
Calcium chloride, anhydrous, powder, 99.99% trace metals basis
Sigma-Aldrich
MPP+ iodide, ≥98% (HPLC), powder
Sigma-Aldrich
Quinidine, crystallized, ≥98.0% (dried material, NT)
Supelco
Calcium ion solution for ISE, 0.1 M Ca, analytical standard (for ion-selective electrodes)
Sigma-Aldrich
Calcium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Ketoconazole, European Pharmacopoeia (EP) Reference Standard
Cimetidine for peak identification, European Pharmacopoeia (EP) Reference Standard
Cimetidine for system suitability, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sulfaphenazole, ≥98% (HPLC)
Sigma-Aldrich
Ketoconazole
Supelco
Ketoconazole, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Cimetidine
Sigma-Aldrich
Nifedipine, ≥98% (HPLC), powder
Supelco
Nifedipine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Quercetin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Quercetin, ≥95% (HPLC), solid
Cimetidine, European Pharmacopoeia (EP) Reference Standard
Nifedipine, European Pharmacopoeia (EP) Reference Standard
Supelco
Cimetidine, Pharmaceutical Secondary Standard; Certified Reference Material