  • Search Results
  • Levosimendan exerts anti-inflammatory effects on cardiac myocytes and endothelial cells in vitro.
Levosimendan exerts anti-inflammatory effects on cardiac myocytes and endothelial cells in vitro.
Thrombosis and haemostasis (2014-10-03)
Konstantin A Krychtiuk, Lukas Watzke, Christoph Kaun, Elisabeth Buchberger, Renate Hofer-Warbinek, Svitlana Demyanets, Julia Pisoni, Stefan P Kastl, Sabine Rauscher, Marion Gröger, Arezu Aliabadi, Andreas Zuckermann, Gerald Maurer, Rainer de Martin, Kurt Huber, Johann Wojta, Walter S Speidl
ABSTRACT
Levosimendan is a positive inotropic drug for the treatment of acute decompensated heart failure (HF). Clinical trials showed that levosimendan was particularly effective in HF due to myocardial infarction. Myocardial necrosis induces a strong inflammatory response, involving chemoattractants guiding polymorphonuclear neutrophils (PMN) into the infarcted myocardial tissue. Our aim was to examine whether levosimendan exhibits anti-inflammatory effects on human adult cardiac myocytes (HACM) and human heart microvascular endothelial cells (HHMEC). Cardiac myocytes and endothelial cells were stimulated with interleukin-1β (IL)-1β (200 U/ml) and treated with levosimendan (0.1-10 µM) for 2-48 hours. IL-1β strongly induced expression of IL-6 and IL-8 in HACM and E-selectin and intercellular adhesion molecule-1 (ICAM-1) in HHMEC and human umbilical vein endothelial cells (HUVEC). Treatment with levosimendan strongly attenuated IL-1β-induced expression of IL-6 and IL-8 in HACM as well as E-selectin and ICAM-1 in ECs. Levosimendan treatment further reduced adhesion of PMN to activated endothelial cells under both static and flow conditions by approximately 50 %. Incubation with 5-hydroxydecanoic acid, a selective blocker of mitochondrial ATP-dependent potassium channels, partly abolished the above seen anti-inflammatory effects. Additionally, levosimendan strongly diminished IL-1β-induced reactive oxygen species and nuclear factor-κB (NF-κB) activity through inhibition of S536 phosphorylation. In conclusion, levosimendan exhibits anti-inflammatory effects on cardiac myocytes and endothelial cells in vitro. These findings could explain, at least in part, the beneficial effects of levosimendan after myocardial infarction.
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Amphotericin B from Streptomyces sp., ~80% (HPLC), powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Amphotericin B from Streptomyces sp., BioReagent, suitable for cell culture, ~80% (HPLC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
SAFC
L-Glutamine
Sigma-Aldrich
Amphotericin B solution, 250 μg/mL in deionized water, 0.1 μm filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Supelco
L-Glutamine, certified reference material, TraceCERT®
Sigma-Aldrich
L-Glutamine
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
HEPES, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
SAFC
HEPES
SAFC
HEPES
Sigma-Aldrich
Levosimendan, ≥98% (HPLC)
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
Amphotericin B solubilized, powder, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)