  • Search Results
  • Validation of a general method for activity estimation of cyanide evolving oxidoreductases.
Validation of a general method for activity estimation of cyanide evolving oxidoreductases.
Analytical biochemistry (2014-12-03)
Francisco Gasteazoro, Ariane Jalila Simaan, Raunel Tinoco-Valencia, Brenda Valderrama
ABSTRACT
Ethylene is a key molecule in organic synthesis currently produced by steam cracking of fossil hydrocarbons. In nature, ethylene is produced in higher plants by 1-aminocyclopropane-1-carboxylic acid oxidase (ACCO). Biocatalytic alternatives for ethylene production are still far from being competitive with traditional production plants. Furthermore, data dispersion shown in the literature adds uncertainty to the introduction of ACCO as a biocatalyst, especially when larger numbers of isoforms or mutants are to be compared. Here we propose a new method for measuring ACCO activity based on cyanide detection. Data provided here indicate that cyanide detection is more precise, more responsive, and much more stable than any other method tested for ACCO activity estimation so far. Briefly, enzymatically produced cyanide can be detected by its derivatization with naphthalene-2,3-dicarboxyaldehide (NDA) to generate 1-cyanobenz[f]isoindole (CBI), which is further detected by high-performance liquid chromatography (HPLC) coupled with a fluorescence detector. Cyanide can be detected in the range between 0.99 and 60.17pmol, which is three orders of magnitude more sensitive than the currently used ethylene estimation method.
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Glycerol, for molecular biology, ≥99.0%
Sigma-Aldrich
Ammonium sulfate, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Ammonium sulfate, BioXtra, ≥99.0%
Sigma-Aldrich
Ammonium sulfate, for molecular biology, ≥99.0%
Sigma-Aldrich
2,2,2-Trichloroethanol, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium bicarbonate-12C, 99.9 atom % 12C
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Decylamine, 95%
Sigma-Aldrich
Ammonium sulfate, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Ammonium-14N2 sulfate, 99.99 atom % 14N
Sigma-Aldrich
Glycerol, FCC, FG
Sigma-Aldrich
Glycerol, BioUltra, for molecular biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Glycerol, tested according to Ph. Eur., anhydrous
Supelco
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Ascorbic acid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Supelco
Decylamine, analytical standard
Sigma-Aldrich
Decylamine, ≥99.0% (GC)
Supelco
Ammonium sulfate, analytical standard, for Nitrogen Determination According to Kjeldahl Method, traceable to NIST SRM 194
Supelco
Ascorbic Acid, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Glycerol, analytical standard
Sigma-Aldrich
Ammonium sulfate, 99.999% trace metals basis
Sigma-Aldrich
Ammonium sulfate, anhydrous, Redi-Dri, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Glycerol, ACS reagent, ≥99.5%
Sigma-Aldrich
Glycerol, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Glycerol, puriss. p.a., ACS reagent, anhydrous, dist., ≥99.5% (GC)
Sigma-Aldrich
Glycerol, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E422, anhydrous, 99.0-101.0% (alkalimetric)