Dietary carotenoids do not improve motility or antioxidant capacity in cichlid fish sperm.
Fish physiology and biochemistry (2014-04-01)
Melissa Sullivan, Alexandria C Brown, Ethan D Clotfelter
RESUMO
Carotenoids may act as antioxidants under many circumstances. We examined the importance of carotenoids as antioxidants in the gonads of male convict cichlids (Amatitlania nigrofasciata), a species in which males lack the carotenoid-based breeding coloration that characterizes females. Male fish were fed one of four diets that included different combinations of xanthophyll and carotene carotenoids, and then we measured carotenoid concentration of the gonads, gonadosomatic index (GSI), sperm motility, and the antioxidant capacity of the gonads. Significant differences were found in gonadal carotenoid content among treatment groups, suggesting that dietary carotenoids were indeed sequestered in the gonads. There were no differences among diet groups, however, in GSI, sperm motility, or gonadal antioxidant capacity. These findings suggest that carotenoids are required only in small amounts in the testes of male convict cichlids or that they play a limited role in protecting sperm from oxidative damage.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Supelco
Ascorbic Acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Supelco
Lutein, analytical standard
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Supelco
L-Ascorbic acid, analytical standard
Supelco
Zeaxanthin, analytical standard
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Ascorbic acid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Supelco
L-Ascorbic acid, certified reference material, TraceCERT®
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
Xanthophyll, from marigold
Sigma-Aldrich
L-Ascorbic acid, tested according to Ph. Eur.
Sigma-Aldrich
L-Ascorbic acid, suitable for plant cell culture