  • Search Results
  • Annexin A2 is implicated in multi-drug-resistance in gastric cancer through p38MAPK and AKT pathway.
Annexin A2 is implicated in multi-drug-resistance in gastric cancer through p38MAPK and AKT pathway.
Neoplasma (2014-08-26)
Z D Zhang, Y Li, Q Fan, B Zhao, B Tan, X F Zhao
ABSTRACT
Studies have shown that Annexin A2 (ANXA2) is related with tumor proliferation, apoptosis, differentiation, invasion, migration, and drug resistance. The purpose of this study was to investigate the role and its mechanisms of ANXA2 in multi-drug-resistance (MDR) in gastric cancer. ANXA2 expression in both gastric cancer tissues and cell lines were detected by quantitative real-time PCR (RT-qPCR) and Western blotting. The cell proliferation was measured by SRB assay. The pool of siRNA against ANXA2 was designed and synthesized and then transfected into resistant gastric cancer SGC7901/DDP cells. ANXA2 expression was detected by RT-qPCR and Western blotting. Drug sensitivities of SGC7901/DDP cells to P-gp-related drug (doxorubicin) and P-gp-non-related drugs (5-FU and cisplatin) were measured by SRB assay. Expression of MDR-related genes and phosphorylation of AKT and MAPKs were also detected by RT-qPCR and Western blotting. Results showed that ANXA2 expression was significantly higher in gastric specimens than that in normal tissues, and negatively correlated with the differentiation level of gastric cancer. In addition, ANXA2 expression level was higher in SGC7901/DDP cells than that in parent SGC7901 cells. After knock-down ANXA2 expression using ANXA2 small interfering RNA, the drug sensitivity of SGC7901/DDP cells to doxorubicin, 5-FU and DDP increased. Delivery of ANXA2 siRNA significantly downregulated the expression of P-gp, MRP1 and Bcl-2, while markedly upregulated Bax in SGC7901/DDP cells. However, several other MDR factors such as GST-π, TOPO-I and TOPO-II had no obvious changes. Additionally, phosphorylation of P38MAPK and AKT, but not ERK1/2 or JNKs was specifically decreased in SGC7901/DDP cells after ANXA2 siRNA delivery. Importantly, P38MAPK and AKT inhibitor increased the drug sensitivity of SGC701/DDP cells in a similar way as ANXA2 siRNAs does. ANXA2 is involved in gastric cancer MDR through regulating p38MAPK and AKT pathways as well as certain MDR factors.
Sigma-Aldrich
β-D-Allose, rare aldohexose sugar
Sigma-Aldrich
DL-Serine, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥98% (HPLC)
Sigma-Aldrich
DL-Serine, ≥98% (TLC)
Sigma-Aldrich
L-Glutathione reduced, BioXtra, ≥98.0%
Sigma-Aldrich
L-Glutathione reduced, suitable for cell culture, BioReagent, ≥98.0%, powder
Sigma-Aldrich
L-Glutathione reduced, ≥98.0%
L-Threonine, European Pharmacopoeia (EP) Reference Standard
Serine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
DL-Glyceraldehyde 3-phosphate solution, 45-55 mg/mL in H2O
Sigma-Aldrich
L-Threonine, reagent grade, ≥98% (HPLC)
Supelco
L-Threonine, certified reference material, TraceCERT®
Sigma-Aldrich
L-Threonine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 99.0-101.0%
SAFC
L-Threonine
Sigma-Aldrich
L-Threonine, BioXtra, ≥99.5% (NT)
Supelco
Glutathione, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Threonine, Pharmaceutical Secondary Standard; Certified Reference Material
Glutathione, European Pharmacopoeia (EP) Reference Standard