  • Search Results
  • Mechanisms of nanoparticle internalization and transport across an intestinal epithelial cell model: effect of size and surface charge.
Mechanisms of nanoparticle internalization and transport across an intestinal epithelial cell model: effect of size and surface charge.
Molecular pharmaceutics (2014-10-21)
Azzah M Bannunah, Driton Vllasaliu, Jennie Lord, Snjezana Stolnik
ABSTRACT
This study investigated the effect of nanoparticle size (50 and 100 nm) and surface charge on their interaction with Caco-2 monolayers as a model of the intestinal epithelium, including cell internalization pathways and the level of transepithelial transport. Initially, toxicity assays showed that cell viability and cell membrane integrity were dependent on the surface charge and applied mass, number, and total surface area of nanoparticles, as tested in two epithelial cell lines, colon carcinoma Caco-2 and airway Calu-3. This also identified suitable nanoparticle concentrations for subsequent cell uptake experiments. Nanoparticle application at doses below half maximal effective concentration (EC₅₀) revealed that the transport efficiency (ratio of transport to cell uptake) across Caco-2 cell monolayers is significantly higher for negatively charged nanoparticles compared to their positively charged counterparts (of similar size), despite the higher level of internalization of positively charged systems. Cell internalization pathways were hence probed using a panel of pharmacological inhibitors aiming to establish whether the discrepancy in transport efficiency is due to different uptake and transport pathways. Vesicular trans-monolayer transport for both positively and negatively charged nanoparticles was confirmed via inhibition of dynamin (by dynasore) and microtubule network (via nocodazole), which significantly reduced the transport of both nanoparticle systems. For positively charged nanoparticles a significant decrease in internalization and transport (46% and 37%, respectively) occurred in the presence of a clathrin pathway inhibitor (chlorpromazine), macropinocytosis inhibition (42%; achieved by 5-(N-ethyl-N-isopropyi)-amiloride), and under cholesterol depletion (38%; via methyl-β-cyclodextrin), but remained unaffected by the inhibition of lipid raft associated uptake (caveolae) by genistein. On the contrary, the most prominent reduction in internalization and transport of negatively charged nanoparticles (51% and 48%, respectively) followed the inhibition of lipid raft-associated pathway (caveolae inhibition by genistein) but was not significantly affected by the inhibition of clathrin pathway.
Sigma-Aldrich
Nocodazole, ≥99% (TLC), powder
Sigma-Aldrich
Genistein, synthetic, ≥98% (HPLC), powder
Sigma-Aldrich
Genistein, from Glycine max (soybean), ~98% (HPLC)
Sigma-Aldrich
Dabco® 33-LV
Sigma-Aldrich
1,4-Diazabicyclo[2.2.2]octane, ReagentPlus®, ≥99%
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Supelco
HEPES, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Genistein, analytical standard
SAFC
HEPES
SAFC
HEPES
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
5-(N-Ethyl-N-isopropyl)amiloride
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture