• Início
  • Resultados da busca
  • Simultaneous quantitative determination of six active components in traditional Chinese medicinal preparation Cerebralcare Granule® by RP-HPLC coupled with diode array detection for quality control.
Simultaneous quantitative determination of six active components in traditional Chinese medicinal preparation Cerebralcare Granule® by RP-HPLC coupled with diode array detection for quality control.
Journal of chromatographic science (2013-08-13)
Xiang-yang Wang, Xiao-hui Ma, Wei Li, Yang Chu, Jia-hua Guo, Shui-ping Zhou, Yong-hong Zhu
RESUMO
A simple, accurate and reliable method for the simultaneous separation and determination of six active components (protocatechuic acid, chlorogenic acid, caffeic acid, paeoniflorin, ferulic acid and rosmarinic acid) in traditional Chinese medicinal preparation Cerebralcare Granule(®) (CG) was developed using reverse-phase high-performance liquid chromatography coupled with diode array detector detection. The chromatographic separation was performed on a Hypersil GOLD C18 column with aqueous formic acid (0.1%, v/v) and acetonitrile as mobile phase at a flow rate of 0.2 ml/min at 30 °C. Because of the different UV characteristics of these components, change detection wavelength method was used for quantitative analysis. All of the analytes showed good linearity (r > 0.9992). The established method showed good precision and relative standard deviations (%) for intra-day and inter-day variations of 0.15-1.81 and 0.11-1.98%, respectively. The validated method was successfully applied to the simultaneously determination of six active components in CG from different batches.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Formic acid, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥98%
Sigma-Aldrich
Acetonitrile, for HPLC, for UV, ≥99.9% (GC)
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Formic acid, puriss., meets analytical specifications of DAC, FCC, 98.0-100%
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile, suitable for HPLC-GC, ≥99.8% (GC)
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Supelco
Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, ≥99.5% (GC)
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, for preparative HPLC, ≥99.8% (GC)
Sigma-Aldrich
Acetonitrile
Sigma-Aldrich
Ultrapure Acetonitrile, for DNA synthesis
Sigma-Aldrich
Acetonitrile, for DNA synthesis
Sigma-Aldrich
Acetonitrile, for DNA synthesis