Merck
  • Início
  • Resultados da busca
  • Combination of bladder cancer-specific oncolytic adenovirus gene therapy with cisplatin on bladder cancer in vitro.
Combination of bladder cancer-specific oncolytic adenovirus gene therapy with cisplatin on bladder cancer in vitro.
Tumour biology : the journal of the International Society for Oncodevelopmental Biology and Medicine (2014-08-03)
Li Wang, Yunxin Zhang, Jinxia Zhao, Erlong Xiao, Jianzhong Lu, Shengjun Fu, Zhiping Wang
RESUMO
Bladder cancer-specific oncolytic adenovirus Ad/PSCAE/UPII/E1A, carrying E1A gene regulated by human Uroplakin II (UPII) promoter and prostate stem cell antigen enhancer (PSCAE), could kill bladder tumor cells preferentially. The aim of this study was to examine the effects of Ad/PSCAE/UPII/E1A combined with cisplatin on human bladder cancer cells and to identify the underlying mechanisms. The combined effects of Ad/PSCAE/UPII/E1A and cisplatin on EJ, 5637, and BIU-87 bladder cancer cells were evaluated by MTT cell proliferation assay. Cell apoptosis was detected by flow cytometry with fluorescein isothiocyanate-conjugated annexin V (annexin V-FITC) and propidium iodide staining. The activation of the caspase pathway and the expression of Bcl-2 family proteins were determined by western blot assay. Ad/PSCAE/UPII/E1A adenovirus vector could infect bladder cancer cell lines selectively and induce growth inhibition effectively. Of note, the combination treatment of cisplatin and Ad/PSCAE/UPII/E1A could inhibit the proliferation of bladder cancer cells significantly compared with the "alone" treatment. Furthermore, Ad/PSCAE/UPII/E1A plus cisplatin combined treatment resulted in enhanced apoptosis in bladder cancer cells. The enhanced antitumor effects in vitro elicited by Ad/PSCAE/UPII/E1A plus cisplatin were closely related to the increased Fas expression and cleavage of caspase-8 and Bid and decrease in the ratio of anti- to pro-apoptotic proteins followed by activation of caspase-9 and caspase-3, which may contribute to the activation of extrinsic and intrinsic apoptotic pathways. Our results indicate that the combination of Ad/PSCAE/UPII/E1A with cisplatin exerts a synergistic antitumor effect on human bladder cancer cells and is a potential combined treatment strategy for bladder cancer.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Adenosine 5′-diphosphoribose sodium salt, ≥93%
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
trans-Platinum(II)diammine dichloride
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Propidium iodide solution, solution (1.0 mg/ml in water)
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
cis-Diammineplatinum(II) dichloride, crystalline
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Cisplatin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
cis-Diamineplatinum(II) dichloride, ≥99.9% trace metals basis
Cisplatin impurity A, European Pharmacopoeia (EP) Reference Standard