  • Search Results
  • An interplay between the p38 MAPK pathway and AUBPs regulates c-fos mRNA stability during mitogenic stimulation.
An interplay between the p38 MAPK pathway and AUBPs regulates c-fos mRNA stability during mitogenic stimulation.
The Biochemical journal (2015-01-15)
Maria Sol Degese, Tamara Tanos, Julian Naipauer, Tim Gingerich, Diego Chiappe, Pablo Echeverria, Jonathan LaMarre, J Silvio Gutkind, Omar A Coso
ABSTRACT
Mitogen-activated protein kinase (MAPK) pathways constitute key regulatory elements linking extracellular stimuli to nuclear gene expression. Immediate-early responsive genes (IEGs) of the activator protein 1 (AP-1) family, such as fos, achieve peak expression levels shortly after cells are stimulated with growth factors and sharply decrease thereafter. Several AU-rich binding proteins (AUBPs), including HuR (Hu-antigen R, Elav-like protein 1, ELAVL1) and KSRP (far upstream element-binding protein 2, KHSRP) bind to a fos AU-rich element (ARE) present in the 3'-UTR (untranslated region) of fos mRNA regulating its stability by a still poorly defined mechanism. We show in the present study that, whereas HuR binds and stabilizes transcribed reporter mRNAs bearing the fos 3'-UTR, KSRP counteracts this effect. Furthermore, we found that fos mRNA stability and HuR phosphorylation status are dependent on the activity of p38 MAPK in both epithelial cells and fibroblasts upon proliferative stimulation. Analysing PPI (protein-protein interaction) networks, we performed a thorough query of interacting proteins for p38 MAPKs, HuR and other AUBPs upon growth factor stimulation. This revealed novel HuR interactors including inhibitors of protein phosphatase 2 (PP2A) activity. Over-expression of two of these interactors, pp32 and APRIL (acidic leucine-rich nuclear phosphoprotein 32 family member B, ANP32B) and pharmacological inhibition of PP2A stabilized a fos reporter mRNA. Our results indicate that p38 MAPK regulates fos mRNA decay by affecting the state of phosphorylation of HuR while controlling yet to be fully elucidated PP regulatory networks.
Sigma-Aldrich
Sigma 7-9®, ≥99% (titration), crystalline
Sigma-Aldrich
Tromethamine, meets USP testing specifications
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Acetonitrile, for DNA synthesis
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Sigma-Aldrich
Fluorescein, for fluorescence, free acid
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
SAFC
L-Glutamine
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Ultrapure Acetonitrile, for DNA synthesis
Millipore
Bifido Selective Supplement B, suitable for microbiology
Sigma-Aldrich
L-Glutamine
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, for HPLC, for UV, ≥99.9% (GC)
Sigma-Aldrich
Formic acid, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥98%
Sigma-Aldrich
Acetonitrile, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%