• Início
  • Resultados da busca
  • Development and validation of determinative and confirmatory LC-MS/MS methodologies for total florfenicol and tulathromycin residues in bovine, equine and porcine kidney, liver and muscle tissues.
Development and validation of determinative and confirmatory LC-MS/MS methodologies for total florfenicol and tulathromycin residues in bovine, equine and porcine kidney, liver and muscle tissues.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2015-01-24)
Rick W Fedeniuk, Del McKenzie, Massey Mizuno, Connie Neiser, Collin O'Byrne, Bryn Shurmer
RESUMO
Separate methods for the quantitation and confirmation of regulatory relevant residue concentrations of total florfenicol and tulathromycin residues in multiple tissue matrices were developed and validated. Total florfenicol residues, determined and expressed as florfenicol amine (FFA) equivalents, were quantified and confirmed over a concentration range of 100-4000ng/g, with an LOD of 33ng/g, while total tulathromyicn residues, determined as CP-60,300 and expressed as tulathromycin equivalents, were quantified and confirmed over a concentration range of 500-10,000ng/g, with an LOD of 300ng/g. A 2 or 1h acid digestion for the FFA and tulathromycin methods, respectively, followed by extraction, cleanup, and concentration using mixed-mode strong cation-exchange SPE cartridges was used. Quantitation and confirmation were accomplished using commercially available tri-deuterated FFA (FFA-D3) as internal standard and multiple reaction monitoring (MRM) of three transitions per target analyte. Mean recoveries and matrix effects were 60% and 25%; and 100% and 110%, respectively for the target analytes florfenicol amine and CP-60,300. Intra-lab method reproducibilities ranged from 7 to 11% RSD, which are within the AOAC recommended HORRATr guidelines for method reproducibilities estimated from single laboratory validation studies. Blind spikes showed that method bias was generally less than 15% for both methods within the calibration range. Both methods have been shown to meet requirements for use in national chemical residue monitoring programs.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
2-Propanol, BioReagent, for molecular biology, ≥99.5%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Supelco
Methanol, analytical standard
Sigma-Aldrich
2-Propanol, suitable for HPLC, 99.9%
Sigma-Aldrich
Ammonium hydroxide solution, ACS reagent, 28.0-30.0% NH3 basis
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Citric acid, ACS reagent, ≥99.5%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
2-Propanol, anhydrous, 99.5%
Sigma-Aldrich
Hexane, suitable for HPLC, ≥97.0% (GC)
Sigma-Aldrich
Hexane, anhydrous, 95%
Sigma-Aldrich
Hexane, ReagentPlus®, ≥99%
Sigma-Aldrich
Hydrogen chloride solution, 4.0 M in dioxane
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Sigma-Aldrich
Citric acid, 99%
Sigma-Aldrich
Ammonium hydroxide solution, 28% NH3 in H2O, ≥99.99% trace metals basis
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
Hydrochloric acid, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., fuming, ≥37%, APHA: ≤10