  • Search Results
  • A label-free fluorescence assay for trypsin based on the electron transfer between oligonucleotide-stabilized Ag nanoclusters and cytochrome c.
A label-free fluorescence assay for trypsin based on the electron transfer between oligonucleotide-stabilized Ag nanoclusters and cytochrome c.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry (2014-08-12)
Mei-Lan Hong, Li-Juan Li, Hui-Xia Han, Xia Chu
ABSTRACT
A label-free fluorescent assay for the detection of trypsin by using oligonucleotide-templated silver nanoclusters (Ag NCs) and cytochrome c (Cyt c) has been demonstrated. When negatively charged Ag NCs and positively charged Cyt c are mixed, they tend to form a hybrid complex, and then lead the fluorescence of Ag NCs to be quenched significantly due to electron transfer between Ag NCs and the heme cofactor of Cyt c. In the presence of trypsin, it catalyzes the hydrolytic cleavage of Cyt c to small peptide fragments, and releases the heme moiety from the Ag NCs/Cyt c complex; the quenched fluorescence restores therewith. By virtue of this specific response, the fluorescent biosensor has a linear range of from 0.7 to 4 μg mL(-1) and from 9 to 120 μg mL(-1) with a detection limit of 58.7 ng mL(-1). Aside from the easy manufacture aspect, our method also possesses a high signal-to-background ratio (~11), excellent selectivity and good biocompatibility, which makes it a promising bioanalysis for a trypsin activity assay.
Sigma-Aldrich
Trizma® base, anhydrous, free-flowing, Redi-Dri, ≥99.9%
Sigma-Aldrich
Trizma® base, BioPerformance Certified, meets EP, USP testing specifications, suitable for cell culture, ≥99.9% (titration)
Sigma-Aldrich
Trizma® base, Primary Standard and Buffer, ≥99.9% (titration), crystalline
Sigma-Aldrich
Trizma® base, ≥99.9% (titration), crystalline
Sigma-Aldrich
Trizma® base, BioXtra, pH 10.5-12.0 (1 M in H2O), ≥99.9% (titration)
Sigma-Aldrich
Trizma® base, puriss. p.a., ≥99.7% (T)
Sigma-Aldrich
Trizma® base, BioUltra, for molecular biology, ≥99.8% (T)
Sigma-Aldrich
Trizma® base, ≥99.0% (T)
Supelco
Trizma® base, certified reference material for titrimetry, certified by BAM, ≥99.5%
Sigma-Aldrich
Tris(hydroxymethyl)aminomethane, ACS reagent, ≥99.8%
Sigma-Aldrich
Tromethamine, meets USP testing specifications
Sigma-Aldrich
Sigma 7-9®, ≥99% (titration), crystalline
SAFC
Tromethamine
Trometamol, European Pharmacopoeia (EP) Reference Standard
Supelco
Tromethamine, pharmaceutical secondary standard, certified reference material