• Início
  • Resultados da busca
  • Preparation and mechanism insight of nuclear envelope-like polymer vesicles for facile loading of biomacromolecules and enhanced biocatalytic activity.
Preparation and mechanism insight of nuclear envelope-like polymer vesicles for facile loading of biomacromolecules and enhanced biocatalytic activity.
ACS nano (2014-06-17)
Yunqing Zhu, Fangyingkai Wang, Cong Zhang, Jianzhong Du
RESUMO
The facile loading of sensitive and fragile biomacromolecules, such as glucose oxidase, hemoglobin, and ribonucleic acid (RNA), via synthetic vehicles directly in pure aqueous media is an important technical challenge. Inspired by the nucleus pore complex that connects the cell nucleus and the cytoplasm across the nuclear envelope, here we describe the development of a kind of polymeric nuclear envelope-like vesicle (NEV) to address this problem. The NEV is tailored to form the polymer pore complex (70 nm, similar to a nucleus pore complex) within the vesicle membrane based on nanophase segregation, which is confirmed via fluorescence spectrometry and dynamic light scattering (DLS) during self-assembly. This pH-triggered polymer pore complex can mediate the transportation of biomacromolecules across the vesicle membrane. Moreover, the NEVs facilitate the natural consecutive enzyme-catalyzed reactions via the H(+) sponge effect. This simple strategy might also be extended for mimicking other synthetic cell organelles.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Triethylamine, ≥99.5%
Sigma-Aldrich
Sodium hydroxide, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Acetone, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Sodium hydroxide solution, 50% in H2O
Sigma-Aldrich
Sodium hydroxide, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Magnesium sulfate, anhydrous, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Triethylamine, ≥99%
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
Sodium hydroxide, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
Acetone, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Potassium carbonate, ACS reagent, ≥99.0%
Sigma-Aldrich
Triethylamine, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Potassium carbonate, 99.995% trace metals basis
Sigma-Aldrich
Triethylamine, BioUltra, ≥99.5% (GC)
Supelco
Sodium hydroxide concentrate, 0.1 M NaOH in water (0.1N), Eluent concentrate for IC
Sigma-Aldrich
Sodium hydroxide, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets
Sigma-Aldrich
4-Methylumbelliferone, ≥98%
Sigma-Aldrich
2-Bromoethanol, 95%
Supelco
Sodium hydroxide solution, 49-51% in water, eluent for IC
Sigma-Aldrich
Acetone, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
Magnesium sulfate, anhydrous, reagent grade, ≥97%
Sigma-Aldrich
Magnesium sulfate, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.8%
Supelco
Acetone, analytical standard
Sigma-Aldrich
N,N,N′,N′′,N′′-Pentamethyldiethylenetriamine, 99%
Sigma-Aldrich
Potassium carbonate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, powder