  • Search Results
  • 4,6-Diaryl/heteroarylpyrimidin-2(1H)-ones as a new class of xanthine oxidase inhibitors.
4,6-Diaryl/heteroarylpyrimidin-2(1H)-ones as a new class of xanthine oxidase inhibitors.
Archiv der Pharmazie (2014-05-02)
Shiwani Shukla, Dinesh Kumar, Ritu Ojha, Manish K Gupta, Kunal Nepali, Preet M S Bedi
ABSTRACT
A series of 4,6-diaryl/heteroarylpyrimidones was synthesized employing silica-supported fluoroboric acid under solvent-free conditions in a microwave reactor. The catalytic influence of HBF4-SiO2 was investigated in detail to optimize the reaction conditions. The synthesized compounds were evaluated for in vitro xanthine oxidase (XO) inhibitory activity for the first time. Structure-activity relationship analyses are also presented. Among the synthesized compounds, VA-5, -9, -10, -12, -22, -23, and -25 were the active inhibitors with IC50 values ranging from 6.45 to 13.46 µM. Compound VA-25 with a pyridinyl ring as ring A and a thiophenyl ring as ring B emerged as the most potent XO inhibitor (IC50 = 6.45 µM) in comparison to allopurinol (IC50 = 12.24 µM). Some of the important interactions of VA-25 with the amino acid residues of the active site of XO were figured out by molecular modeling studies.
Sigma-Aldrich
Ammonium-14N2 sulfate solution, 40 wt. % in H2O, 99.99 atom % 14N
Sigma-Aldrich
Ammonium-14N2,sulfate-16O4, 99.99 atom % 14N, 99.99 atom % 16O
Sigma-Aldrich
Ammonium sulfate-14N2 solution, 40 wt. % in H2O, 99.99 atom % 14N
Sigma-Aldrich
Ammonium sulfate, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Ammonium-14N2 sulfate, 99.99 atom % 14N
Sigma-Aldrich
Uric acid, ≥99%, crystalline
Sigma-Aldrich
Uric acid, BioXtra, ≥99% (HPLC)
Sigma-Aldrich
Ammonium sulfate, for molecular biology, ≥99.0%
Sigma-Aldrich
Ammonium sulfate, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Ammonium sulfate, BioXtra, ≥99.0%
Sigma-Aldrich
Ammonium sulfate, 99.999% trace metals basis
Supelco
Ammonium sulfate, analytical standard, for Nitrogen Determination According to Kjeldahl Method, traceable to NIST SRM 194
Sigma-Aldrich
Ammonium sulfate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.0%
Sigma-Aldrich
Ammonium sulfate, anhydrous, Redi-Dri, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Ammonium sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Ammonium sulfate, ReagentPlus®, ≥99.0%