Merck
Specific increase in potency via structure-based design of a TCR.
Journal of immunology (Baltimore, Md. : 1950) (2014-07-30)
Karolina Malecek, Arsen Grigoryan, Shi Zhong, Wei Jun Gu, Laura A Johnson, Steven A Rosenberg, Timothy Cardozo, Michelle Krogsgaard
ABSTRACT
Adoptive immunotherapy with Ag-specific T lymphocytes is a powerful strategy for cancer treatment. However, most tumor Ags are nonreactive "self" proteins, which presents an immunotherapy design challenge. Recent studies have shown that tumor-specific TCRs can be transduced into normal PBLs, which persist after transfer in ∼30% of patients and effectively destroy tumor cells in vivo. Although encouraging, the limited clinical responses underscore the need for enrichment of T cells with desirable antitumor capabilities prior to patient transfer. In this study, we used structure-based design to predict point mutations of a TCR (DMF5) that enhance its binding affinity for an agonist tumor Ag-MHC (peptide-MHC [pMHC]), Mart-1 (27L)-HLA-A2, which elicits full T cell activation to trigger immune responses. We analyzed the effects of selected TCR point mutations on T cell activation potency and analyzed cross-reactivity with related Ags. Our results showed that the mutated TCRs had improved T cell activation potency while retaining a high degree of specificity. Such affinity-optimized TCRs have demonstrated to be very specific for Mart-1 (27L), the epitope for which they were structurally designed. Although of somewhat limited clinical relevance, these studies open the possibility for future structural-based studies that could potentially be used in adoptive immunotherapy to treat melanoma while avoiding adverse autoimmunity-derived effects.
Sigma-Aldrich
Interleukin-2 human, IL-2, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Sigma-Aldrich
β-D-Allose, rare aldohexose sugar
Sigma-Aldrich
Interleukin-2 human, IL-2, recombinant, expressed in HEK 293 cells, suitable for cell culture, endotoxin tested
Sigma-Aldrich
Interleukin-2 human, recombinant, expressed in Pichia pastoris, suitable for cell culture
Sigma-Aldrich
IL-2 human, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Sigma-Aldrich
IL-2 human, recombinant, expressed in HEK 293 cells, ≥95% (SDS-PAGE)
Sigma-Aldrich
Chromium, powder, 99.5%, −100 mesh
Sigma-Aldrich
Chromium, chips, thickness ~2 mm, 99.5%
Sigma-Aldrich
Interleukin-2 human, recombinant, expressed in E. coli, ~10000 U/mL
Sigma-Aldrich
Chromium, powder, ≥99% trace metals basis, <45 μm
Sigma-Aldrich
Chromium, chips, 99.995% trace metals basis
Chromium, foil, light tested, 25x25mm, thickness 0.03mm, permanent polyester support, 99.99+%
Chromium, foil, light tested, 25x25mm, thickness 0.04mm, permanent polyester support, 99.99+%
Chromium, foil, light tested, 25x25mm, thickness 0.025mm, permanent polyester support, 99.99+%
Chromium, foil, 50x50mm, thickness 3.0mm, hard, 99.95%
Chromium, foil, not light tested, 50x50mm, thickness 0.025mm, permanent polyester support, 99.99+%
Chromium, foil, not light tested, 50x50mm, thickness 0.02mm, permanent polyester support, 99.99+%
Chromium, foil, not light tested, 50x50mm, thickness 0.03mm, permanent polyester support, 99.99+%
Chromium, foil, not light tested, 50x50mm, thickness 0.04mm, permanent polyester support, 99.99+%
Chromium, microfoil, disks, 10mm, thinness 1.0μm, specific density 719μg/cm2, permanent mylar 3.5μm support, 99.99+%
Chromium, microfoil, disks, 25mm, thinness 0.05μm, specific density 37.27μg/cm2, permanent mylar 3.5μm support, 99.99+%
Chromium, microfoil, disks, 25mm, thinness 0.25μm, specific density 178.7μg/cm2, permanent mylar 3.5μm support, 99.99+%
Chromium, microfoil, disks, 25mm, thinness 0.5μm, specific density 355μg/cm2, permanent mylar 3.5μm support, 99.99+%
Chromium, microfoil, disks, 25mm, thinness 1.0μm, specific density 719μg/cm2, permanent mylar 3.5μm support, 99.99+%
Chromium, rod, 100mm, diameter 2.0mm, 99.7+%
Chromium, foil, not light tested, 100x100mm, thickness 0.0125mm, permanent polyester support, 99.99+%
Chromium, foil, not light tested, 100x100mm, thickness 0.015mm, permanent polyester support, 99.99+%
Chromium, foil, not light tested, 100x100mm, thickness 0.01mm, permanent polyester support, 99.99+%
Chromium, foil, not light tested, 100x100mm, thickness 0.025mm, permanent polyester support, 99.99+%
Chromium, foil, not light tested, 100x100mm, thickness 0.02mm, permanent polyester support, 99.99+%