  • Search Results
  • A switch in the source of ATP production and a loss in capacity to perform glycolysis are hallmarks of hepatocyte failure in advance liver disease.
A switch in the source of ATP production and a loss in capacity to perform glycolysis are hallmarks of hepatocyte failure in advance liver disease.
Journal of hepatology (2014-03-04)
Taichiro Nishikawa, Nadège Bellance, Aaron Damm, Han Bing, Zhen Zhu, Kan Handa, Mladen I Yovchev, Vasudha Sehgal, Tyler J Moss, Michael Oertel, Prahlad T Ram, Iraklis I Pipinos, Alejandro Soto-Gutierrez, Ira J Fox, Deepak Nagrath
ABSTRACT
The cause of hepatic failure in the terminal stages of chronic injury is unknown. Cellular metabolic adaptations in response to the microenvironment have been implicated in cellular breakdown. To address the role of energy metabolism in this process we studied mitochondrial number, respiration, and functional reserve, as well as cellular adenosine-5'-triphosphate (ATP) production, glycolytic flux, and expression of glycolysis related genes in isolated hepatocytes from early and terminal stages of cirrhosis using a model that produces hepatic failure from irreversible cirrhosis in rats. To study the clinical relevance of energy metabolism in terminal stages of chronic liver failure, we analyzed glycolysis and energy metabolism related gene expression in liver tissue from patients at different stages of chronic liver failure according to Child-Pugh classification. Additionally, to determine whether the expression of these genes in early-stage cirrhosis (Child-Pugh Class A) is related to patient outcome, we performed network analysis of publicly available microarray data obtained from biopsies of 216 patients with hepatitis C-related Child-Pugh A cirrhosis who were prospectively followed up for a median of 10years. In the early phase of cirrhosis, mitochondrial function and ATP generation are maintained by increasing energy production from glycolytic flux as production from oxidative phosphorylation falls. At the terminal stage of hepatic injury, mitochondria respiration and ATP production are significantly compromised, as the hepatocytes are unable to sustain the increased demand for high levels of ATP generation from glycolysis. This impairment corresponds to a decrease in glucose-6-phosphatase catalytic subunit and phosphoglucomutase 1. Similar decreased gene expression was observed in liver tissue from patients at different stages of chronic liver injury. Further, unbiased network analysis of microarray data revealed that expression of these genes was down regulated in the group of patients with poor outcome. An adaptive metabolic shift, from generating energy predominantly from oxidative phosphorylation to glycolysis, allows maintenance of energy homeostasis during early stages of liver injury, but leads to hepatocyte dysfunction during terminal stages of chronic liver disease because hepatocytes are unable to sustain high levels of energy production from glycolysis.
Sigma-Aldrich
Sucrose, Grade I, suitable for plant cell culture
Supelco
Sucrose, analytical standard, for enzymatic assay kit SCA20
Sigma-Aldrich
Sucrose, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ACS reagent
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, Grade II, suitable for plant cell culture
Supelco
Sucrose, analytical standard
Sigma-Aldrich
Sucrose, ≥99.5%
Sigma-Aldrich
Sucrose, meets USP testing specifications
Sigma-Aldrich
Sucrose, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, BioUltra, for molecular biology, ≥99.5% (HPLC)
Millipore
Sucrose, ACS reagent, suitable for microbiology, ≥99.0%
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Sucrose, European Pharmacopoeia (EP) Reference Standard
Supelco
Sucrose, Pharmaceutical Secondary Standard; Certified Reference Material