  • Search Results
  • Correlation among the BRAF gene mutation status, clinicopathological features of primary tumour, and lymph node metastasizing of papillary thyroid carcinoma.
Correlation among the BRAF gene mutation status, clinicopathological features of primary tumour, and lymph node metastasizing of papillary thyroid carcinoma.
Experimental and clinical endocrinology & diabetes : official journal, German Society of Endocrinology [and] German Diabetes Association (2014-05-20)
J Lukas, J Drabek, B Dudesek, P Vazan, J Stranska, S Jancik, M Mackova, M Syrucek, D Lukas, J Duskova, P Dundr, B Hintnausova, J Jiskra
ABSTRACT
Papillary thyroid carcinoma (PTC) is the most common malignant thyroid tumour. A common mutation of papillary thyroid carcinoma (PTC) is the somatic mutation of the BRAF (V600E) gene. The aim was to 1) determine the association of lymph node metastases of PTC with the BRAF gene mutation of primary tumour; 2) evaluate association of the BRAF mutation in the -primary tumour with clinicopathological para-meters; 3) examine the extent of genetic heterogeneity by monitoring the BRAF mutation in multicentric tumours. Retrospective analysis of the BRAF (V600E) mutation in PTC and PTC neck lymph node metastases in 156 patients operated from 2003 to 2012 in Prague and Zlín, the Czech Republic, using a qPCR assay. The results were correlated with clinicopathological factors. DNA was successfully extracted from 137 samples. The BRAF (V600E) mutation was detected in 78 cases (56.9%). The patients with BRAF p.Val600Glu mutation of primary tumour had only non-significantly higher risk of cervical lymph node metastases [OR=2.39 (95%) CI 1.00-5.75, p=0.052]. In the classic papillary variant, the BRAF (V600E) mutation was found significantly more often than in other PTC subtypes (p=0.022). We did not confirm any significant association between the BRAF (V600E) mutation and other clinicopathological findings. Except for the higher prevalence in papillary variant of PTC, BRAF p.Val600Glu mutation was not associated with other prognostic clinicopathological factors of PTC. BRAF mutation cannot be regarded as a reliable marker of node metastases in patients with PTC.
Sigma-Aldrich
Ethanol, purum, secunda spirit, denaturated with 2% 2-butanone and 0.5% 4-methyl-2-pentanone, S15, ~96% (based on denaturant-free substance)
Supelco
Ethanol, standard for GC
Supelco
o-Xylene, analytical standard
Sigma-Aldrich
o-Xylene, reagent grade, ≥98.0%
Sigma-Aldrich
Ethanol, purum, absolute ethanol, denaturated with 4.8% isopropanol, A15 IPA1, ≥99.8% (based on denaturant-free substance)
Sigma-Aldrich
Ethanol, ACS reagent, prima fine spirit, without additive, F15 o1
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 2% 2-butanone, F25 MEK1, ~96% (based on denaturant-free substance)
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
Sigma-Aldrich
Ethanol, purum, absolute ethanol, denaturated with 1% cyclohexane, A15 CYCLO1, ≥99.8% (based on denaturant-free substance)
Sigma-Aldrich
Ethanol, purum, absolute ethanol, denaturated with 2% 2-butanone, A15 MEK1, ≥99.8% (based on denaturant-free substance)
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, HPLC/spectrophotometric grade
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
o-Xylene, puriss. p.a., ≥99.0% (GC)
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, for molecular biology
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, for molecular biology
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS reagent, meets USP testing specifications, Excise Tax-free, Permit for use required
Sigma-Aldrich
Ethyl alcohol, Pure, 160 proof, Excise Tax-free, Permit for use required
Supelco
o-Xylene, suitable for HPLC, 98%
Sigma-Aldrich
o-Xylene, anhydrous, 97%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Supelco
o-Xylene, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethanol, BioUltra, for molecular biology, ≥99.8%, (absolute alcohol, without additive, A15 o1)
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol