  • Search Results
  • Cytoprotective effect of eckol against oxidative stress-induced mitochondrial dysfunction: involvement of the FoxO3a/AMPK pathway.
Cytoprotective effect of eckol against oxidative stress-induced mitochondrial dysfunction: involvement of the FoxO3a/AMPK pathway.
Journal of cellular biochemistry (2014-04-05)
Areum Daseul Kim, Kyoung Ah Kang, Mei Jing Piao, Ki Cheon Kim, Jian Zheng, Cheng Wen Yao, Ji Won Cha, Chang Lim Hyun, Hee Kyoung Kang, Nam Ho Lee, Jin Won Hyun
ABSTRACT
This study investigated the cytoprotective effect of Ecklonia cava-derived eckol against H2O2-induced mitochondrial dysfunction in Chang liver cells. While H2O2 augmented levels of mitochondrial reactive oxygen species (ROS), eckol decreased it. Eckol also attenuated high intracellular Ca(2+) levels stimulated by H2O2 and recovered H2O2-diminished ATP levels and succinate dehydrogenase activity. Eckol time-dependently increased the expression of manganese superoxide dismutase (Mn SOD), a mitochondrial antioxidant enzyme with cytoprotective effect against oxidative stress. Eckol recovered Mn SOD expression and activity that were decreased by H2O2. Finally, eckol induced Mn SOD through phosphorylated AMP-activated protein kinase (AMPK) and forkhead box O3a (FoxO3a). Specific silencing RNAs (siRNAs) against FoxO3a and AMPK reduced eckol-stimulated Mn SOD expression, and diethyldithiocarbamate (Mn SOD inhibitor) and siRNA against Mn SOD reduced the cytoprotective effect of eckol against H2O2-provoked cell death. These results demonstrate that eckol protects cells from mitochondrial oxidative stress by activating AMPK/FoxO3a-mediated induction of Mn SOD.
Sigma-Aldrich
DL-Serine, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥98% (HPLC)
Sigma-Aldrich
DL-Serine, ≥98% (TLC)
L-Threonine, European Pharmacopoeia (EP) Reference Standard
Serine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dihydrorhodamine 123, ≥95%
Sigma-Aldrich
L-Threonine, reagent grade, ≥98% (HPLC)
Supelco
L-Threonine, certified reference material, TraceCERT®
Sigma-Aldrich
L-Threonine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 99.0-101.0%
SAFC
L-Threonine
Sigma-Aldrich
L-Threonine, BioXtra, ≥99.5% (NT)
Supelco
L-Threonine, Pharmaceutical Secondary Standard; Certified Reference Material