• Início
  • Resultados da busca
  • Regulatory T cell levels and cytokine production in active non-infectious uveitis: in-vitro effects of pharmacological treatment.
Regulatory T cell levels and cytokine production in active non-infectious uveitis: in-vitro effects of pharmacological treatment.
Clinical and experimental immunology (2014-10-31)
B Molins, M Mesquida, R W J Lee, V Llorenç, L Pelegrín, A Adán
RESUMO
The aim of this study was to quantify the proportion of regulatory T cells (Treg ) and cytokine expression by peripheral blood mononuclear cells (PBMCs) in patients with active non-infectious uveitis, and to evaluate the effect of in-vitro treatment with infliximab, dexamethasone and cyclosporin A on Treg levels and cytokine production in PBMCs from uveitis patients and healthy subjects. We included a group of 21 patients with active non-infectious uveitis and 18 age-matched healthy subjects. The proportion of forkhead box protein 3 (FoxP3)(+) Treg cells and intracellular tumour necrosis factor (TNF)-α expression in CD4(+) T cells was determined by flow cytometry. PBMCs were also either rested or activated with anti-CD3/anti-CD28 and cultured in the presence or absence of dexamethasone, cyclosporin A and infliximab. Supernatants of cultured PBMCs were collected and TNF-α, interleukin (IL)-10, IL-17 and interferon (IFN)-γ levels were measured by enzyme-linked immunosorbent assay (ELISA). No significant differences were observed in nTreg levels between uveitis patients and healthy subjects. However, PBMCs from uveitis patients produced significantly higher amounts of TNF-α and lower amounts of IL-10. Dexamethasone treatment in vitro significantly reduced FoxP3(+) Treg levels in PBMCs from both healthy subjects and uveitis patients, and all tested drugs significantly reduced TNF-α production in PBMCs. Dexamethasone and cyclosporin A significantly reduced IL-17 and IFN-γ production in PBMCs and dexamethasone up-regulated IL-10 production in activated PBMCs from healthy subjects. Our results suggest that PBMCs from patients with uveitis express more TNF-α and less IL-10 than healthy subjects, and this is independent of FoxP3(+) Treg levels. Treatment with infliximab, dexamethasone and cyclosporin A in vitro modulates cytokine production, but does not increase the proportion of FoxP3(+) Treg cells.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, ≥99% (TLC), film or powder
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
Dexamethasone, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
Ionomycin calcium salt from Streptomyces conglobatus, powder, ≥98% (HPLC)
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
Dexamethasone, ≥98% (HPLC), powder
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, suitable for protein labeling, ≥90% (HPLC), powder
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Ionomycin calcium salt, Ready Made Solution, from Streptomyces conglobatus, 1 mM in DMSO
SAFC
HEPES
SAFC
L-Glutamine
Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
Supelco
Dexamethasone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
Sigma-Aldrich
Dexamethasone, powder, γ-irradiated, BioXtra, suitable for cell culture, ≥80% (HPLC)
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, synthetic, ≥98.0% (TLC)
Sigma-Aldrich
Aphidicolin from Nigrospora sphaerica, ≥98% (HPLC), powder
SAFC
HEPES
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Dexamethasone, meets USP testing specifications
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Dexamethasone, European Pharmacopoeia (EP) Reference Standard
Supelco
L-Glutamine, certified reference material, TraceCERT®