  • Search Results
  • Intestinal myofibroblast TRPC6 channel may contribute to stenotic fibrosis in Crohn's disease.
Intestinal myofibroblast TRPC6 channel may contribute to stenotic fibrosis in Crohn's disease.
Inflammatory bowel diseases (2015-02-04)
Lin Hai Kurahara, Miho Sumiyoshi, Kunihiko Aoyagi, Keizo Hiraishi, Kyoko Nakajima, Midori Nakagawa, Yaopeng Hu, Ryuji Inoue
ABSTRACT
Intestinal fibrosis is a frequent complication of Crohn's disease (CD) and often leads to detrimental stricture formation. Myofibroblasts play active roles in mediating fibrotic changes in various tissues. We investigated whether transient receptor potential channels in intestinal myofibroblasts are involved in CD-associated intestinal fibrosis. An intestinal myofibroblast cell line (InMyoFibs) was stimulated with transforming growth factor-β1 (TGF-β1) to model excessive fibrosis. Biopsy samples from nonstenotic or stenotic intestinal regions from patients with CD were used for quantitative comparisons of transient receptor potential channel and fibrosis-associated factor expression levels. TGF-β1 treatment transformed spindle-shaped InMyoFibs into filament-shaped cells with enhanced α-actin stress fiber formation, transient receptor potential canonical (TRPC) 4 and TRPC6 messenger RNA and protein expression, and basal- and agonist-induced Ca influxes. TGF-β1 also enhanced the formation of TRPC6/smooth muscle α-actin, TRPC6/N-cadherin, and TRPC4/N-cadherin coimmunoprecipitates. Inhibition of TRPC6 in InMyoFibs by RNA interference or dominant-negative mutations suppressed TGF-β1-induced Ca influxes, stress fiber formation, and smooth muscle α-actin expression, but increased COL1A1, interleukin (IL)-10, and IL-11 expression, as well as Smad-2, ERK, and p38-MAPK phosphorylation. Similar increases in phosphorylation levels were observed with TRPC and calcineurin inhibitors. In stenotic areas in patients with CD, TRPC6, ACTA2 (smooth muscle α-actin), CDH2 (N-cadherin), COL1A1, IL-10, and IL-11 were significantly increased. These results suggest that augmented Ca influxes due to TRPC6 upregulation facilitate stress fiber formation and strengthen cell-cell interactions by negatively regulating the synthesis of antifibrotic factors in TGF-β1-treated myofibroblasts. Similar changes observed in stenotic areas of patients with CD suggest the therapeutic significance of targeting TRPC6.
Sigma-Aldrich
Sigma 7-9®, ≥99% (titration), crystalline
Sigma-Aldrich
Tromethamine, meets USP testing specifications
Supelco
Cyclosporine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Tromethamine, pharmaceutical secondary standard, certified reference material
SAFC
Tromethamine
Trometamol, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Trizma® base, Primary Standard and Buffer, ≥99.9% (titration), crystalline
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Cyclosporin A, from Tolypocladium inflatum, ≥95% (HPLC), solid
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Carbamoylcholine chloride, ≥98% (titration), crystalline
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Tris(hydroxymethyl)aminomethane, ACS reagent, ≥99.8%
Sigma-Aldrich
Trizma® base, ≥99.9% (titration), crystalline
Sigma-Aldrich
Trizma® base, BioXtra, pH 10.5-12.0 (1 M in H2O), ≥99.9% (titration)
Sigma-Aldrich
Trizma® base, BioPerformance Certified, meets EP, USP testing specifications, suitable for cell culture, ≥99.9% (titration)
Supelco
Picric acid solution, 100 μg/mL in acetonitrile, PESTANAL®, analytical standard
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Picric acid solution, 1.3% in H2O (saturated)
Sigma-Aldrich
Cyclosporin A, 97.0-101.5% (on dried basis)
Supelco
Sodium chloride, certified reference material for titrimetry, certified by BAM, ≥99.5%
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Supelco
Trizma® base, certified reference material for titrimetry, certified by BAM, ≥99.5%
Sigma-Aldrich
Trizma® base, puriss. p.a., ≥99.7% (T)
Sigma-Aldrich
Trizma® base, ≥99.0% (T)