• Início
  • Resultados da busca
  • Screening of drugs of abuse and toxic compounds in human whole blood using online solid-phase extraction and high-performance liquid chromatography with time-of-flight mass spectrometry.
Screening of drugs of abuse and toxic compounds in human whole blood using online solid-phase extraction and high-performance liquid chromatography with time-of-flight mass spectrometry.
Journal of separation science (2014-10-29)
Xiaomei Teng, Chen Liang, Rong Wang, Tao Sun, Yulan Rao, Chunfang Ni, Libo Zeng, Lingjuan Xiong, Yuan Li, Yurong Zhang
RESUMO
A novel method for the screening of 151 drugs of abuse and toxic compounds in human whole blood has been developed and validated by online solid-phase extraction with liquid chromatography coupled to time-of-flight mass spectrometry. Analytes were extracted and separated by using a fully automated online solid-phase extraction liquid chromatography system with total chromatographic run time of 26 min. Time-of-flight mass spectrometry screening of 151 drugs of abuse and toxic compounds was performed in a full-scan (m/z 50-800) mode using an MS(E) acquisition of molecular ions and fragment ions data at two collision energies (one was 6 eV and another one was in the range of 5-45 eV). The compounds were identified based on retention times and exact mass of molecular ions and fragment ions. The limit of detection ranged from 1 to 100 ng/mL and the recovery of the method ranged from 6.3 to 163.5%. This method is proved to be a valuable screening method allowing fast and specific identification of drugs in human whole blood.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Supelco
Methanol, analytical standard
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Supelco
Ammonium formate, eluent additive for LC-MS, LiChropur, ≥99.0%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Formic acid, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥98%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Acetonitrile, for HPLC, for UV, ≥99.9% (GC)
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Ammonium formate, reagent grade, 97%
Sigma-Aldrich
Ammonium formate, ≥99.995% trace metals basis
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Formic acid, puriss., meets analytical specifications of DAC, FCC, 98.0-100%
Sigma-Aldrich
Ammonium formate, BioUltra, ≥99.0% (calc. based on dry substance, NT)
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ammonium formate solution, BioUltra, 10 M in H2O
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for HPLC-GC, ≥99.8% (GC)