Merck
  • Início
  • Resultados da busca
  • Angiotensin II acting on brain AT1 receptors induces adrenaline secretion and pressor responses in the rat.
Angiotensin II acting on brain AT1 receptors induces adrenaline secretion and pressor responses in the rat.
Scientific reports (2014-11-29)
Kumiko Nakamura, Takahiro Shimizu, Toshihiko Yanagita, Takayuki Nemoto, Keisuke Taniuchi, Shogo Shimizu, Fotios Dimitriadis, Toshio Yawata, Youichirou Higashi, Tetsuya Ueba, Motoaki Saito
RESUMO
Angiotensin II (AngII) plays important roles in the regulation of cardiovascular function. Both peripheral and central actions of AngII are involved in this regulation, but mechanisms of the latter actions as a neurotransmitter/neuromodulator within the brain are still unclear. Here we show that (1) intracerebroventricularly (i.c.v.) administered AngII in urethane-anesthetized male rats elevates plasma adrenaline derived from the adrenal medulla but not noradrenaline with valsartan- (AT1 receptor blocker) sensitive brain mechanisms, (2) peripheral AT1 receptors are not involved in the AngII-induced elevation of plasma adrenaline, although AngII induces both noradrenaline and adrenaline secretion from bovine adrenal medulla cells, and (3) i.c.v. administered AngII elevates blood pressure but not heart rate with the valsartan-sensitive mechanisms. From these results, i.c.v. administered AngII acts on brain AT1 receptors, thereby inducing the secretion of adrenaline and pressor responses. We propose that the central angiotensinergic system can activate central adrenomedullary outflow and modulate blood pressure.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Aluminum oxide, mesoporous, MSU-X (wormhole), average pore size 3.8 nm
Cytarabine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Butyric acid, ≥99%, FG
Sigma-Aldrich
Butyric acid, natural, ≥99%, FCC, FG
Sigma-Aldrich
Aluminum oxide, nanowires, diam. × L 2-6 nm × 200-400 nm
Sigma-Aldrich
Aluminum oxide, 99.997% trace metals basis
Supelco
Aluminum oxide, activated, neutral, Brockmann Activity I
Supelco
Butyric acid, analytical standard
Supelco
Aluminum oxide, for the determination of hydrocarbons
Sigma-Aldrich
Aluminum oxide, pellets, 3 mm
Sigma-Aldrich
Aluminum oxide, fused, powder, primarily α-phase, 100-200 mesh
Sigma-Aldrich
Aluminum oxide, Corundum, α-phase, -100 mesh
Sigma-Aldrich
Aluminum oxide, fused, powder, primarily α-phase, -325 mesh
Sigma-Aldrich
Aluminum oxide, calcined, powder, primarily α-phase, 100-325 mesh
Sigma-Aldrich
Aluminum oxide, activated, acidic, Brockmann I, free-flowing, Redi-Dri
Sigma-Aldrich
Aluminum oxide, activated, neutral, Brockmann I, free-flowing, Redi-Dri
Sigma-Aldrich
Aluminum oxide, activated, basic, Brockmann I
Sigma-Aldrich
Aluminum oxide, activated, acidic, Brockmann I
Sigma-Aldrich
Aluminum oxide, activated, neutral, Brockmann I
Sigma-Aldrich
Aluminum oxide, Type WN-6, Neutral, Activity Grade Super I
Sigma-Aldrich
Aluminum oxide, powder, primarily α phase, ≤10 μm avg. part. size, 99.5% trace metals basis
Sigma-Aldrich
Aluminum oxide, pore size 58 Å, ~150 mesh
Sigma-Aldrich
Aluminum oxide, powder, 99.99% trace metals basis
Sigma-Aldrich
Aluminum oxide, nanoparticles, <50 nm particle size (DLS), 20 wt. % in isopropanol
Sigma-Aldrich
Aluminum oxide, nanoparticles, 30-60 nm particle size (TEM), 20 wt. % in H2O
Sigma-Aldrich
Aluminum oxide, nanopowder, 13 nm primary particle size (TEM), 99.8% trace metals basis
Sigma-Aldrich
Aluminum oxide, nanopowder, <50 nm particle size (TEM)
Sigma-Aldrich
Aluminum oxide, single crystal substrate, <0001>
Sigma-Aldrich
Butyric acid, ≥99%