Downregulation of Tryptophan-related Metabolomic Profile in Rheumatoid Arthritis Synovial Fluid.
The Journal of rheumatology (2015-09-04)
Kwi Young Kang, Soo Hyun Lee, Seung Min Jung, Sung-Hwan Park, Byung-Hwa Jung, Ji Hyeon Ju
RESUMO
Synovial fluid (SF) is one of the most important materials that reflect the pathophysiological process of arthritis. A metabolomic and lipidomic study of SF was performed with the aim of identifying tentative diagnostic markers or therapeutic candidates for rheumatoid arthritis (RA). SF was aspirated from 10 patients with RA and 10 patients with osteoarthritis (OA). RA SF and OA SF were collected and analyzed by ultraperformance liquid chromatography quadruple time-of-flight mass spectrometry. Associations among clinical variables, laboratory results, and metabolic profiles were investigated. The metabolic pathways for carnitine, tryptophan, phenylalanine, arachidonic acid, and glycophospholipid were significantly upregulated in OA SF. The metabolic pathways for taurine, cholesterol ester, and the β-oxidation of pristine acid, linolenic acid, and sphingolipid were activated more in RA SF than in OA SF. In particular, the tryptophan pathway, which comprises kynurenine, indoleacetic acid, indole acetaldehyde, and N'-formylkynurenine, was downregulated. Interestingly, the levels of tryptophan metabolites kynurenine and N'-formylkynurenine, which are involved in immune tolerance, were significantly lower in RA SF compared with OA SF (p < 0.05), but the opposite pattern was observed for erythrocyte sedimentation rate (p < 0.01) and the levels of C-reactive protein (CRP; p < 0.01), rheumatoid factor (p < 0.01), and anticyclic citrullinated peptide antibody (p < 0.05). Kynurenine concentration correlated inversely with CRP concentration in RA SF but not in OA SF (r -0.65, p < 0.05). Advances in metabolomic techniques enabled us to delineate distinctive metabolic and lipidomic profiles in RA SF and OA SF. RA SF and OA SF showed distinct metabolic profiles.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Caffeine, powder, ReagentPlus®
Sigma-Aldrich
Formic acid, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥98%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Adipic acid, 99%
Sigma-Aldrich
Acetaminophen, BioXtra, ≥99.0%
Sigma-Aldrich
Caffeine, anhydrous, 99%, FCC, FG
Sigma-Aldrich
Formic acid, puriss., meets analytical specifications of DAC, FCC, 98.0-100%
Sigma-Aldrich
Acetaminophen, meets USP testing specifications, 98.0-102.0%, powder
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Acetaminophen, analytical standard
Sigma-Aldrich
Sodium formate, ACS reagent, ≥99.0%
Sigma-Aldrich
Caffeine, Sigma Reference Standard, vial of 250 mg
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Hippuric acid, 98%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Sodium formate, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
Giemsa stain, certified by the Biological Stain Commission
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Caffeine, meets USP testing specifications, anhydrous
Sigma-Aldrich
Adipic acid, BioXtra, ≥99.5% (HPLC)
Sigma-Aldrich
Sodium formate, reagent grade, 97%
Sigma-Aldrich
Reserpine