  • Search Results
  • Expression, purification and identification of Pla a1 in a codon-optimized Platanus pollen allergen.
Expression, purification and identification of Pla a1 in a codon-optimized Platanus pollen allergen.
Molecular medicine reports (2015-04-23)
Yun Liu, Xiuzhen Sun, Guizuo Wang, Ailin Tao, Yuanyuan Wu, Manxiang Li, Hongyang Shi, Mei Xie
ABSTRACT
The present study aimed to express, purify and identify the major allergen gene, Pla a1, in Platanus pollen. According to previous studies, the major gene sequences of the Pla a1 allergen were obtained and codon optimization and synthesis of the genome were performed using DNAStar software. Following binding of the target gene fragment and the pET-44a vector, the JM109 cells were transfected to produce positive clones. The vectors were then transformed into Escherichia coli Rosetta cells to induce the expression of the target protein. The exogenous protein was purified using affinity chromatography and was identified by western blot analysis. Pla a1, the major allergen protein in Platanus pollen, was successfully isolated and this exogenous protein was purified using affinity chromatography. The present study was the first, to the best of our knowledge, to obtain expression of the allergen recombinant protein, Pla a1, fused with a Strep-TagII via codon optimization and provided the basis for the preparation of allergens with high purity, recombinant hypoallergenic allergens and allergen nucleic acid vaccines.
Sigma-Aldrich
IPTG, ≥99% (TLC), ≤0.1% Dioxane
Sigma-Aldrich
Potassium phosphate monobasic, powder, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Potassium phosphate monobasic, ReagentPlus®
Sigma-Aldrich
Potassium phosphate monobasic, for molecular biology, ≥98.0%
Sigma-Aldrich
Isopropyl β-D-1-thiogalactopyranoside, ≥99% (TLC)
SAFC
Isopropyl β-D-1-thiogalactopyranoside
Sigma-Aldrich
Potassium phosphate monobasic, BioUltra, for molecular biology, anhydrous, ≥99.5% (T)
Sigma-Aldrich
Isopropyl β-D-thiogalactopyranoside solution, ReadyMade IPTG solution for Blue-white screening
Sigma-Aldrich
Potassium phosphate monobasic, 99.99% trace metals basis
Sigma-Aldrich
L-Thyroxine sodium salt pentahydrate, ≥98% (HPLC), powder
Sigma-Aldrich
L-Thyroxine sodium salt pentahydrate, γ-irradiated, powder, BioXtra, suitable for cell culture