  • Search Results
  • An electrochemical microRNAs biosensor with the signal amplification of alkaline phosphatase and electrochemical-chemical-chemical redox cycling.
An electrochemical microRNAs biosensor with the signal amplification of alkaline phosphatase and electrochemical-chemical-chemical redox cycling.
Analytica chimica acta (2015-05-24)
Ning Xia, Youjuan Zhang, Xin Wei, Yaping Huang, Lin Liu
ABSTRACT
MicroRNAs (MiRNAs) have been regarded as clinically important biomarkers and drug discovery targets. In this work, we reported a simple and ultrasensitive electrochemical method for miRNAs detection based on single enzyme amplification and electrochemical-chemical-chemical (ECC) redox cycling. Specifically, upon contact with the target miRNAs, the hairpin structure of biotinylated DNA immobilized on gold electrode was destroyed and the biotin group in DNA was forced away from the electrode surface, allowing for the coupling of streptavidin-conjugated alkaline phosphatase (SA-ALP). Then, ascorbic acid (AA, the enzymatic product of ALP) triggered the ECC redox cycling with ferrocene methanol (FcM) and tris(2-carboxyethyl)phosphine (TCEP) as the redox mediator and the chemical reducing reagent, respectively. The method was more sensitive than that with horseradish peroxidase (HRP) or glucose oxidase (GOx) triggered recycling since one ALP molecule captured by one target miRNA molecule promoted the production of thousands of AA. Analytical merits (e.g., detection limit, dynamic range, specificity, regeneration and reproducibility) were evaluated. The feasibility of the method for analysis of miRNA-21 in human serum has also been demonstrated.
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
1-Hexanethiol, 95%
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
6-Mercapto-1-hexanol, 97%
Sigma-Aldrich
Ferrocenemethanol, 97%
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
L-Ascorbic acid, suitable for plant cell culture
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Sigma-Aldrich
6-Mercapto-1-hexanol, 99%
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
1-Hexanethiol, 99% (GC)
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC