  • Search Results
  • N-glycosylation profile analysis of Trastuzumab biosimilar candidates by Normal Phase Liquid Chromatography and MALDI-TOF MS approaches.
N-glycosylation profile analysis of Trastuzumab biosimilar candidates by Normal Phase Liquid Chromatography and MALDI-TOF MS approaches.
Journal of proteomics (2015-04-25)
Ivan Sanchez-De Melo, Paola Grassi, Francisco Ochoa, Jorge Bolivar, Francisco J García-Cózar, Ma Carmen Durán-Ruiz
ABSTRACT
The pharmaceutical market has entered an era in which the production of new therapeutics is being often replaced by "biosimilars", copies of already commercialized products waiting for the patents to expire in order to be distributed in a more competitive and affordable manners. Due to its relevance, the ErbB2-targeted monoclonal antibody Trastuzumab (Herceptin) used as breast cancer therapy is one of the main targets in the production of biosimilars. A major challenge is to produce antibodies with the same or the closest N-glycosylation pattern seen in the commercialized drug. Several factors, such as growing conditions or cell types employed, can determine the final composition and structure of the glycans, significantly affecting the properties of the generated antibodies. Therefore, an appropriate characterization is essential. In the present study, we describe two different but complementary strategies to characterize the N-glycosylation of two biosimilar candidates of Trastuzumab. In the first case, N-glycans are fluorescently labeled and separated by Normal Phase HPLC. Different sugars will elute at different times and can be identified using specific oligosaccharide standards. In the second approach, released glycans are permethylated and analyzed by MALDI-TOF MS, being able to determine the structure because of the differential sugar masses. The characterization of the N-glycosylation sites of therapeutic recombinant monoclonal antibodies (mAbs) is usually one of the most critical and time consuming steps in the developing process of biosimilars or any other glycosylated drug. Herein we describe two different but complementary approaches to characterize mAbs glycosylation patterns, the use of glycan fluorescence labeling coupled to HPLC and MALDI-TOF MS profile analysis. This article is part of a Special Issue entitled: HUPO 2014.
Sigma-Aldrich
Sigma 7-9®, ≥99% (titration), crystalline
Sigma-Aldrich
Tromethamine, meets USP testing specifications
Sigma-Aldrich
Indole-3-acetic acid sodium salt, BioReagent, suitable for plant cell culture, ≥98%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
2,5-Dihydroxybenzoic acid, 98%
Supelco
2,5-Dihydroxybenzoic acid, matrix substance for MALDI-MS, >99.0% (HPLC)
Sigma-Aldrich
Acetonitrile, for DNA synthesis
Supelco
Iodomethane solution, certified reference material, 2000 μg/mL in methanol: water (4:1)
Sigma-Aldrich
Acetonitrile, for DNA synthesis
Sigma-Aldrich
Ultrapure Acetonitrile, for DNA synthesis
Sigma-Aldrich
Acetonitrile
Sigma-Aldrich
Ultrapure Acetonitrile
SAFC
Tromethamine
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, ≥99.5% (GC)
Sigma-Aldrich
Acetonitrile, for preparative HPLC, ≥99.8% (GC)
Sigma-Aldrich
Ammonium bicarbonate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Trizma® base, Primary Standard and Buffer, ≥99.9% (titration), crystalline
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Tris(hydroxymethyl)aminomethane, ACS reagent, ≥99.8%