Preparation and Characterization of a Monoclonal Antibody Against Morphine.
Monoclonal antibodies in immunodiagnosis and immunotherapy (2015-08-25)
Susan Kashanian, Ali Shams, Hossein Ghahremani, Maliheh Paknejad
RESUMO
A monoclonal antibody (MAb) was produced by immunization of a BALB/c mouse with a conjugated morphine C6-hemisuccinated derivative (MHS) to cationized bovine serum albumin (cBSA). The hybridoma clones were screened by indirect ELISA using MHS-BSA. The best hybridoma clone was subcloned thrice by limiting dilution. This hybridoma was found to be of IgG2b class and subclass and contained lambda light chain. The affinity of the MAb to morphine was obtained 2.8×10(9) M(-1). The titer of the cell culture supernatant was at least 1:800. The MAb was cross-reacted with codeine (100%) and apomorphine (16.5%), but not with heroin, naloxone, naltrexone, or papaverine. Morphine was conjugated to HRP using a mixed anhydride method and a direct competitive ELISA was designed using anti-morphine MAb. The assay was sensitive over the 50 ng/mL to 5 μg/mL concentration range. In conclusion, this MAb is useful for the development of immunoassays to measure morphine in urine.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Triethylamine, ≥99.5%
Sigma-Aldrich
Triethylamine, ≥99%
Sigma-Aldrich
1,2-Dichloroethane, anhydrous, 99.8%
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Sigma-Aldrich
Triethylamine, puriss. p.a., ≥99.5% (GC)
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Triethylamine, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
L-Lysine monohydrochloride, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
Isobutyl chloroformate, 98%
Sigma-Aldrich
L-Lysine monohydrochloride, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Ethylenediamine dihydrochloride, 98%
Sigma-Aldrich
Triethylamine, for amino acid analysis, ≥99.5% (GC)
Sigma-Aldrich
L-Lysine monohydrochloride, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
Triethylamine, for protein sequence analysis, ampule, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, ≥99.5%