• Início
  • Resultados da busca
  • Injectable and photocross-linkable gels based on gellan gum methacrylate: a new tool for biomedical application.
Injectable and photocross-linkable gels based on gellan gum methacrylate: a new tool for biomedical application.
International journal of biological macromolecules (2014-12-03)
Settimio Pacelli, Patrizia Paolicelli, Inge Dreesen, Shuichiro Kobayashi, Annabella Vitalone, Maria Antonietta Casadei
RESUMO
In this work, a natural polysaccharide gellan gum (GG) has been modified with methacrylic groups (GG-MA) and combined with polyethylene glycol dimethacrylate (PEG-DMA) in order to create novel injectable hydrogels that can be easily delivered through a needle and photocross-linked in the injection site. A novel synthetic procedure for methacrylation of GG has been proposed to better control its derivatization. Different degrees of functionalization have been achieved and their effects on the solubility and mechanical properties of GG-MA were investigated. A good balance in terms of hydrophilicity and elasticity of the corresponding hydrogels was identified, although not suitable enough as injectable material for the treatment of damaged soft tissues. For this reason, several concentrations and different molecular weights of PEG-DMA were investigated to modulate the composition of GG-MA hydrogels and overcome their extreme fragility. Swelling abilities of the hydrogels in different media were studied as a key parameter able to affect the release profile of loaded therapeutic agents. Model molecules having different spherical hindrance (sulindac and vitamin B12) were then chosen to study how the hydrogels were able to modulate their diffusion profiles over time. Finally, the hydrogel's safety was evaluated trough an MTT cytotoxicity test on human fibroblasts.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Triethylamine, ≥99.5%
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
1-Methyl-2-pyrrolidinone, anhydrous, 99.5%
Sigma-Aldrich
Diethyl ether, contains 1 ppm BHT as inhibitor, anhydrous, ≥99.7%
Sigma-Aldrich
Triethylamine, ≥99%
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
4-(Dimethylamino)pyridine, ReagentPlus®, ≥99%
Sigma-Aldrich
Vitamin B12, ≥98%
Sigma-Aldrich
Triethylamine, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
Streptomycin sulfate salt, powder
Sigma-Aldrich
Methacrylic anhydride, contains 2,000 ppm topanol A as inhibitor, 94%
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Triethylamine, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Streptomycin sulfate salt, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Vitamin B12, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Deuterium oxide, 99 atom % D
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D, contains 0.05 wt. % 3-(trimethylsilyl)propionic-2,2,3,3-d4 acid, sodium salt
SAFC
L-Glutamine
Sigma-Aldrich
Deuterium oxide, "100%", ≥99.96 atom % D
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)