• Início
  • Resultados da busca
  • Candidate biomarkers for mosquito age-grading identified by label-free quantitative analysis of protein expression in Aedes albopictus females.
Candidate biomarkers for mosquito age-grading identified by label-free quantitative analysis of protein expression in Aedes albopictus females.
Journal of proteomics (2015-08-15)
I Iovinella, B Caputo, E Michelucci, F R Dani, A della Torre
RESUMO
We applied a "shotgun" approach based on nanoliquid chromatography-high resolution mass spectrometry associated to label free quantification (LFQ) to identify proteins varying with age, independently from the physiological state, in Aedes albopictus, a mosquito species which in the last decades invaded temperate regions in North America and Europe, creating concerns for associated high nuisance and risk of arbovirus transmission. The combined "shotgun" and LFQ approach was shown to be highly suitable to simultaneously compare several biological samples, as needed in a study aimed to analyze different age-groups and physiological states of adult mosquito females. The results obtained represent the first wide-scale analysis of protein expression in Ae. albopictus females: >1000 and 665 proteins were identified from few micrograms of crude protein extracts of mosquito heads and thoraxes, respectively. Six of these proteins were shown to significantly vary from 2- to 16-day-old females, independently from their physiological state (i.e. virgin, mated, host-seeking, blood-fed, and gravid). Mosquito-borne diseases, such as malaria, dengue and other arboviroses, are a persistent cause of global mortality and morbidity, affecting hundreds of thousands of people. Billions of people living in tropical areas are at risk of being bitten every day by an infective mosquito female and the spread of tropical species such as Aedes albopictus to temperate areas is creating alarm in the northern hemisphere. Mosquito longevity is a critical factor affecting mosquito-borne pathogen transmission cycles and the mosquito capacity to transmit pathogens. However, large scale analyses of the age structure of mosquito field populations is hampered by the lack of optimal age-grading approaches. Our findings open new perspectives for the development of reliable, simple and cheap protein-based assays to age-grade Ae. albopictus females and, most likely, other mosquito species of higher medical relevance, such as the main dengue vector, Aedes aegypti, and the major Afrotropical malaria vectors. These assays would greatly contribute to epidemiological studies aimed at defining the actual vectorial capacity of a given mosquito species. Moreover, they would be very valuable in assessing the effectiveness of mosquito control interventions based on the relative ratio between young and old individuals before and after the intervention.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Trifluoroacetic acid, ReagentPlus®, 99%
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Urea, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Urea, powder, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Iodoacetamide, BioUltra
Sigma-Aldrich
Ammonium bicarbonate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Sigma-Aldrich
Formic acid, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥98%
Sigma-Aldrich
Thiourea, ACS reagent, ≥99.0%
Sigma-Aldrich
Acetic acid, glacial, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, 99.8-100.5%
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Iodoacetamide, Single use vial of 56 mg
Sigma-Aldrich
Trifluoroacetic acid, puriss. p.a., suitable for HPLC, ≥99.0% (GC)
Sigma-Aldrich
Iodoacetamide, ≥99% (NMR), crystalline
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Urea, ReagentPlus®, ≥99.5%, pellets
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Formic acid, puriss., meets analytical specifications of DAC, FCC, 98.0-100%