  • Search Results
  • Rac1 GTPase-deficient HeLa cells present reduced DNA repair, proliferation, and survival under UV or gamma irradiation.
Rac1 GTPase-deficient HeLa cells present reduced DNA repair, proliferation, and survival under UV or gamma irradiation.
Molecular and cellular biochemistry (2015-03-12)
Gisele Espinha, Juliana H Osaki, Yuli T Magalhaes, Fabio Luis Forti
ABSTRACT
Rac1 GTPase controls essential cellular functions related to the cytoskeleton, such as motility and adhesion. Rac1 is overexpressed in many tumor cells, including breast cancers, where it is also involved in the proliferation and checkpoint control necessary for the cell's recovery after exposure to ionizing radiation. However, its role in DNA damage and repair remains obscure in other tumor cells and under different genotoxic conditions. Here, we compare HeLa cells with mutants exogenously expressing a dominant-negative Rac1 (HeLa-Rac1-N17) by their responses to DNA damage induced by gamma or UV radiation. In HeLa cells, these treatments led to increased levels of active Rac1 (Rac1-GTP) and of stress fibers, with a diminished ability to migrate compared to untreated cells. However, the reduction of Rac1-GTP in Rac1-N17-deficient clones resulted in much higher levels of polymerized stress fibers accompanied by a strong impairment of cell migration, even after both radiation treatments. With regard to proliferation and genomic stability, dominant-negative Rac1 cells were more sensitive to gamma and UV radiation, exhibiting reduced proliferation and survival consistent with increased DNA damage and delayed or reduced DNA repair observed in this Rac1-deficient clone. The DNA damage response, as indicated by pH2AX and pChk1 levels, was increased in HeLa cells but was not effectively triggered in the Rac1-N17 clone after radiation treatment, which is likely the main cause of DNA damage accumulation. These data suggest that Rac1 GTPase plays an important role in signaling and contributes to the sensitivity of cervical cancer cells under UV or gamma radiation treatments.
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
L-Glutathione reduced, ≥98.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
IPTG, ≥99% (TLC), ≤0.1% Dioxane
Sigma-Aldrich
Sodium deoxycholate, ≥97% (titration)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Magnesium chloride solution, for molecular biology, 1.00 M±0.01 M
Sigma-Aldrich
Magnesium chloride, anhydrous, ≥98%
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Sodium deoxycholate, BioXtra, ≥98.0% (dry matter, NT)
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
L-Glutathione reduced, suitable for cell culture, BioReagent, ≥98.0%, powder
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
Isopropyl β-D-1-thiogalactopyranoside, ≥99% (TLC)
SAFC
Sodium deoxycholate