  • Search Results
  • Inhibition of spawning in zebrafish (Danio rerio): Adverse outcome pathways of quinacrine and ethinylestradiol.
Inhibition of spawning in zebrafish (Danio rerio): Adverse outcome pathways of quinacrine and ethinylestradiol.
General and comparative endocrinology (2015-02-04)
Madelyne M Cosme, Andrea L Lister, Glen Van Der Kraak
ABSTRACT
This study determined the effects of the estrogen receptor agonist ethinylestradiol (EE2) and the phospholipase A2 inhibitor quinacrine (QUIN) on the pathways controlling follicular development, steroidogenesis, oocyte maturation, ovulation and spawning success in adult zebrafish. Both EE2 and QUIN inhibited spawning but did so through different mechanisms. EE2 affected follicular development (reduced ovarian size and reduction in the proportion of cortical alveolus, vitellogenic and mature follicle stages), steroidogenesis (reduced expression of aromatase), maturation (reduced luteinizing hormone receptor expression) and ovulation (reduced expression of cytosolic phospholipase A2 and the nuclear progesterone receptor). Although EE2 alters the proportion of follicle stages within the ovary, the downregulation of gene expression as a consequence of EE2 exposure was primarily due to a decline in expression of the genes of interest in vitellogenic and mature ovarian follicles. QUIN targeted ovulation via a reduction of the steroid 17α,20β dihydroxy-4-prenen-3-one (17α,20β-P) and decreased expression of the prostaglandin metabolizing enzyme cyclooxygenase 2. This study demonstrates the usefulness in defining the impacts of toxicants at the molecular and cellular, organ and whole organism level and how connections between these impacts can be used to describe the adverse outcome pathways (AOPs) that mediate toxicant action. Histological analysis and gene expression were effective tools in defining the AOPs of QUIN and EE2 while the measurement of reproductive hormones level did not provide much valuable information regarding the toxicant's mode of action.
Product Description
Supelco
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
Sodium azide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium azide, BioXtra
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Butyl methyl ether, 99%
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
L-Ascorbic acid, suitable for plant cell culture
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Sigma-Aldrich
Methanol, NMR reference standard