  • Search Results
  • Stabilization of Chromobacterium viscosum Lipase (CVL) Against Ultrasound Inactivation by the Pretreatment with Polyethylene Glycol (PEG).
Stabilization of Chromobacterium viscosum Lipase (CVL) Against Ultrasound Inactivation by the Pretreatment with Polyethylene Glycol (PEG).
Applied biochemistry and biotechnology (2015-09-17)
Md Mahabubur Rahman Talukder, Simon Choo Sze Shiong
ABSTRACT
Although ultrasound has been used to accelerate many enzymatic reactions, the low stability of enzymes in such a system still remains a critical issue, limiting its industrial application. Here, we have reported that polyethylene glycol (PEG) pretreatment stabilized Chromobacterium viscosum lipase (CVL) in ultrasound-assisted water-isooctane emulsion. PEGs of different molecular weights and concentrations were used to pretreat CVL, and the pretreated lipase activities for olive oil hydrolysis were investigated at different ultrasonic powers. The best result was attained with PEG400 at 100 mg/ml for a lipase concentration of 0.02 mg/ml and an ultrasonic power of 106 W. The half-life time of PEG400-treated lipase at 106 W was 54 min, a 27-fold higher than that attained using untreated lipase. Circular dichroism (CD) spectra suggested that PEG increased the rigidity of CVL structure, which favored the lipase stability against ultrasound inactivation. These results have important implications for the exploitation of ultrasound in biocatalytic process.
Sigma-Aldrich
Oleic acid, suitable for cell culture, BioReagent
Sigma-Aldrich
Oleic acid, meets analytical specification of Ph, Eur., 65.0-88.0% (GC)
Sigma-Aldrich
2,2,4-Trimethylpentane, anhydrous, 99.8%
Sigma-Aldrich
Oleic acid, technical grade, 90%
Sigma-Aldrich
Oleic acid, ≥99% (GC)
Sigma-Aldrich
Oleic acid, natural, FCC
Sigma-Aldrich
2,2,4-Trimethylpentane, puriss. p.a., ACS reagent, ≥99.5% (GC)
Sigma-Aldrich
2,2,4-Trimethylpentane, ReagentPlus®, ≥99%
Sigma-Aldrich
2,2,4-Trimethylpentane, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
2,2,4-Trimethylpentane, ACS reagent, ≥99.0%