Merck
Inhibition of SERPINe1 reduces rhabdoviral infections in zebrafish.
Fish & shellfish immunology (2015-09-13)
Amparo Estepa, Julio Coll
ABSTRACT
While exploring the molecular mechanisms behind the fin hemorrhages that follow zebrafish (Danio rerio) early infection with viral haemorrhagic septicemia virus (VHSV), we discovered that most serpin (serine protease inhibitor) gene transcripts were upregulated, except those of serpine1. Surprisingly, only SERPINe1-derived 14-mer peptide and low molecular weight drugs targeting SERPINe1 (i.e. tannic acid, EGCG, tiplaxtinin) inhibited in vitro infections not only of VHSV, but also of other fish rhabdoviruses such as infectious hematopoietic necrosis virus (IHNV) and spring viremia carp virus (SVCV). While the mechanisms that inhibited rhabdoviral infections remain speculative, these and other results suggested that SERPINEe1-derived peptide specifically targeted viral infectivity rather than virions. Practical applications might be developed from these studies since preliminary evidences showed that tannic acid could be used to reduce VHSV-caused mortalities. These studies are an example of how the identification of host genes targeted by viral infections using microarrays might facilitate the identification of novel prevention drugs in aquaculture and illuminate viral infection mechanisms.
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
SAFC
HEPES
SAFC
HEPES
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
Amphotericin B solubilized, powder, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
DL-Serine, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥98% (HPLC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Dicyanocobinamide, ≥93%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Amphotericin B from Streptomyces sp., ~80% (HPLC), powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
DL-Serine, ≥98% (TLC)
Sigma-Aldrich
Amphotericin B from Streptomyces sp., BioReagent, suitable for cell culture, ~80% (HPLC)
Sigma-Aldrich
Amphotericin B solution, 250 μg/mL in deionized water, 0.1 μm filtered, BioReagent, suitable for cell culture