  • Search Results
  • Developmentally regulated expression and expression strategies of Drosophila snoRNAs.
Developmentally regulated expression and expression strategies of Drosophila snoRNAs.
Insect biochemistry and molecular biology (2015-02-03)
Alberto Angrisani, Hakim Tafer, Peter F Stadler, Maria Furia
ABSTRACT
Small nucleolar RNAs constitute a significant portion of the eukaryotic small ncRNA transcriptome and guide site-specific methylation or pseudouridylation of target RNAs. In addition, they can play diverse regulatory roles on gene expression, acting as precursors of smaller fragments able to modulate alternative splicing or operate as microRNAs. Defining their expression strategies and the full repertory of their biological functions is a critical, but still ongoing, process in most organisms. Considering that Drosophila melanogaster is one of the most advantageous model organism for genetic, functional and developmental studies, we analysed the whole genomic organization of its annotated snoRNAs - whose vast majority is known to be embedded in an intronic context - and show by GO term enrichment analysis that protein-coding genes involved in cell division and cytoskeleton organization are those mostly preferred as hosts. This finding was unexpected, and delineates an unpredicted link between snoRNA host genes and cell proliferation that might be of general relevance. We also defined by quantitative RT-PCR the expression of a representative subset of annotated specimens throughout the life cycle, providing a first overview on developmental profiling of the fly snoRNA transcriptome. We found that most of the tested specimens, rather than acting as housekeeping genes with uniform expression, exhibit dynamic developmental expression patterns; moreover, intronic snoRNAs harboured by the same host gene often exhibit distinct temporal profiles, indicating that they can be expressed uncoordinatedly. In addition to provide an updated outline of the fly snoRNA transcriptome, our data highlight that expression of these versatile ncRNAs can be finely regulated.
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
Liquified Phenol, ≥89.0%
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Phenol solution, Equilibrated with 10 mM Tris HCl, pH 8.0, 1 mM EDTA, BioReagent, for molecular biology
Sigma-Aldrich
Phenol solution, Saturated with 0.1 M citrate buffer, pH 4.3 ± 0.2, BioReagent, for molecular biology
Sigma-Aldrich
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (GC)
Sigma-Aldrich
Phenol, BioUltra, for molecular biology, TE-saturated, ~73% (T)
Sigma-Aldrich
Phenol, for molecular biology
Sigma-Aldrich
Phenol, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
Phenol, ≥99%
Sigma-Aldrich
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, ≥99.5% (GC), crystalline (detached)
Sigma-Aldrich
Phenol, ≥96.0% (calc. on dry substance, T)
Sigma-Aldrich
Phenol, puriss. p.a., ACS reagent, reag. Ph. Eur., 99.0-100.5%
Sigma-Aldrich
Phenol, unstabilized, ReagentPlus®, ≥99%
Sigma-Aldrich
Phenol, unstabilized, purified by redistillation, ≥99%
Sigma-Aldrich
Phenol, contains hypophosphorous as stabilizer, loose crystals, ACS reagent, ≥99.0%
Sigma-Aldrich
Phenol, natural, 97%, FG