  • Search Results
  • Ethylene degreening modulates health promoting phytochemicals in Rio Red grapefruit.
Ethylene degreening modulates health promoting phytochemicals in Rio Red grapefruit.
Food chemistry (2015-06-05)
Priyanka R Chaudhary, G K Jayaprakasha, Bhimanagouda S Patil
ABSTRACT
In the current study, we examined the effects of postharvest degreening and storage on phytochemicals in Rio Red grapefruit. Grapefruits were degreened with 3.5 μl/l of ethylene at 21 °C and 80% relative humidity for 72 h, while non-degreened fruits were used as the control. Furthermore, the grapefruits were stored at 11 °C for 3 weeks and then at 21 °C for 2 weeks. Degreening improved the peel colour of the grapefruit without affecting total soluble solids or acidity of the juice. Degreened fruits had significantly more ascorbic acid after 35 days of storage. Degreening had no significant effect on the levels of carotenoids, limonoids and flavonoids as compared to the non-degreened fruits, after 35 days of storage. However, after 7 days, degreened fruits had more limonin and flavonoids and less furocoumarin, namely 6',7'-dihydroxybergamottin. Overall, ethylene treatment had a significant effect on the phytochemical contents of Rio Red grapefruit, especially after 7 days of storage.
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
Limonin, from citrus seeds, >90% (HPLC)
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
Lycopene, ≥90%, from tomato
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Lycopene, Redivivo, 10% FS, ~10% in corn oil, ≥95.0% (sum of isomers)
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
L-Ascorbic acid, suitable for plant cell culture
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
Butylated hydroxytoluene, ≥99%, FCC, FG
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
2,6-Di-tert-butyl-4-methylphenol, ≥99.0% (GC), powder
Sigma-Aldrich
Naringin, ≥90%, from citrus fruit
Supelco
2,5-Dihydroxybenzoic acid, matrix substance for MALDI-MS, >99.0% (HPLC)
Sigma-Aldrich
Naringin, ≥95% (HPLC)
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Sigma-Aldrich
2,5-Dihydroxybenzoic acid, 98%