Merck
  • Início
  • Resultados da busca
  • Simulation of Observed PCBs and Pesticides in the Water Column during the North Atlantic Bloom Experiment.
Simulation of Observed PCBs and Pesticides in the Water Column during the North Atlantic Bloom Experiment.
Environmental science & technology (2015-06-09)
Lin Zhang, Louis Thibodeaux, Lee Jones, Rainer Lohmann
RESUMO
The dynamics of persistent organic pollutants in the oceans are not well constrained, in particular during a bloom formation and collapse. Polychlorinated biphenyls (PCBs) and some pesticides were measured in air, water, and zooplankton tracking the North Atlantic Bloom in May 2008. Lower weight PCBs were entering the water column from the atmosphere during the main bloom period but reached equilibrium after the bloom collapsed. The PCBs in the lipids of zooplankton Calanus were in equilibrium with those in the dissolved phase. A Lagrangian box model was developed to simulate the dissolved phase PCBs and pesticides by including the following processes: air-water exchange, reversible sorption to POC, changes in mixed layer depth, removal by sinking particles, and degradation. Results suggest that sorption to (sinking) POC was the dominant removal process for hydrophobic pollutants from seawater. Statistical test suggested simulated results were not significantly different from observed values for hydrophobic pollutants (p,p'-DDE).
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Aluminum oxide, mesoporous, MSU-X (wormhole), average pore size 3.8 nm
Sigma-Aldrich
Aluminum oxide, nanopowder, 13 nm primary particle size (TEM), 99.8% trace metals basis
Sigma-Aldrich
Aluminum oxide, nanoparticles, <50 nm particle size (DLS), 20 wt. % in isopropanol
Sigma-Aldrich
Aluminum oxide, 99.997% trace metals basis
Sigma-Aldrich
Aluminum oxide, nanowires, diam. × L 2-6 nm × 200-400 nm
Sigma-Aldrich
Aluminum oxide, nanopowder, <50 nm particle size (TEM)
Sigma-Aldrich
Aluminum oxide, single crystal substrate, <0001>
Sigma-Aldrich
Aluminum oxide, activated, acidic, Brockmann I
Sigma-Aldrich
Aluminum oxide, activated, basic, Brockmann I
Sigma-Aldrich
Aluminum oxide, activated, neutral, Brockmann I
Sigma-Aldrich
Aluminum oxide, activated, acidic, Brockmann I, free-flowing, Redi-Dri
Sigma-Aldrich
Aluminum oxide, activated, neutral, Brockmann I, free-flowing, Redi-Dri
Sigma-Aldrich
Aluminum oxide, pellets, 3 mm
Sigma-Aldrich
Aluminum oxide, pore size 58 Å, ~150 mesh
Sigma-Aldrich
Aluminum oxide, Type WN-6, Neutral, Activity Grade Super I
Sigma-Aldrich
Aluminum oxide, powder, primarily α phase, ≤10 μm avg. part. size, 99.5% trace metals basis
Sigma-Aldrich
Aluminum oxide, powder, 99.99% trace metals basis
Sigma-Aldrich
Aluminum oxide, fused, powder, primarily α-phase, -325 mesh
Sigma-Aldrich
Aluminum oxide, calcined, powder, primarily α-phase, 100-325 mesh
Sigma-Aldrich
Aluminum oxide, fused, powder, primarily α-phase, 100-200 mesh
Sigma-Aldrich
Aluminum oxide, Corundum, α-phase, -100 mesh
Sigma-Aldrich
Aluminum oxide, nanoparticles, 30-60 nm particle size (TEM), 20 wt. % in H2O