  • Search Results
  • Circulating microparticles from patients with valvular heart disease and cardiac surgery inhibit endothelium-dependent vasodilation.
Circulating microparticles from patients with valvular heart disease and cardiac surgery inhibit endothelium-dependent vasodilation.
The Journal of thoracic and cardiovascular surgery (2015-07-07)
Li Fu, Xiao-Xia Hu, Ze-Bang Lin, Feng-Jun Chang, Zhi-Jun Ou, Zhi-Ping Wang, Jing-Song Ou
ABSTRACT
Vascular function is very important for maintaining circulation after cardiac surgery. Circulating microparticles (MPs) generated in various diseases play important roles in causing inflammation, coagulation, and vascular injury. However, the impact of MPs generated from patients who have valvular heart disease (VHD), before and after cardiac surgery, on vascular function remains unknown. This study is designed to investigate the impact of such MPs on vasodilation. Microparticles were isolated from age-matched healthy subjects and patients who had VHD, before cardiac surgery, and at 12 hours and 72 hours afterward. The number of MPs was measured and compared. Effects evaluated were of the impact of MPs on: vasodilation of mice aorta; the phosphorylation and expression of Akt, endothelial nitric oxide synthase (eNOS), protein kinase C-βII (PKC-βII), and p70 ribosomal protein S6 kinase (p70S6K); expression of caveolin-1; the association of eNOS with heat shock protein 90 (HSP90); and generation of nitric oxide and superoxide anion of human umbilical vein endothelial cells. Compared with the healthy subjects, VHD patients had significantly higher levels of circulating MPs and those MPs before cardiac surgery can: impair endothelium-dependent vasodilation; inhibit phosphorylation of Akt and eNOS; increase activation of PKC-βII and p70S6K; enhance expression of caveolin-1; reduce the association of HSP90 with eNOS; decrease nitric oxide production, and increase superoxide anion generation. These deleterious effects were even stronger in postoperative MPs. Our data demonstrate that MPs generated from VHD patients before and after cardiac surgery contributed to endothelial dysfunction, by uncoupling and inhibiting eNOS. Circulating MPs are potential therapeutic targets for the maintenance of vascular function postoperatively.
Sigma-Aldrich
DL-Serine, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥98% (HPLC)
Sigma-Aldrich
DL-Serine, ≥98% (TLC)
Sigma-Aldrich
DL-Glyceraldehyde 3-phosphate solution, 45-55 mg/mL in H2O
Sigma-Aldrich
(−)-Norepinephrine, ≥98%, crystalline