• Início
  • Resultados da busca
  • Plasma pharmacokinetics of the indenoisoquinoline topoisomerase I inhibitor, NSC 743400, in rats and dogs.
Plasma pharmacokinetics of the indenoisoquinoline topoisomerase I inhibitor, NSC 743400, in rats and dogs.
Cancer chemotherapy and pharmacology (2015-03-18)
Miguel Muzzio, Shu-Chieh Hu, Julianne L Holleran, Robert A Parise, Julie L Eiseman, Archibong E Yellow-Duke, Joseph M Covey, Elizabeth R Glaze, Kory Engelke, Merrill J Egorin, David L McCormick, Jan H Beumer
RESUMO
NSC 743400 is a novel synthetic indenoisoquinoline analog under development as an anticancer agent. It is a potent topoisomerase I inhibitor with potential therapeutic advantages over FDA-approved camptothecin derivatives. In preparation for clinical development of NSC 743400, we determined the pharmacokinetics after administration to rats and dogs. NSC 743400 was administered intravenously at a dose of 12 or 24 mg/m(2) to rats (single bolus) or 10, 50, 100, 215, 430, or 646 mg/m(2) (intravenous infusion) or 860 or 1720 mg/m(2) (orally) to dogs. Intravenously administered NSC 743400 was eliminated from both species with an estimated t 1/2 of 2-5 h in rat and 6-14 h in dog. Elimination t 1/2 increased with dose in dog. Area under the plasma concentration-versus-time curve (AUC) was comparable in both species, at about 300-400 h ng/mL for the approximately 10 mg/m(2) dose groups. Overall, AUC values increased proportionally with dose for both species but had evidence of more than proportional exposure at the highest doses. Oral dosing resulted in variable drug absorption. The pharmacokinetic data were used to plan first-in-human clinical trials.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
Citric acid, ACS reagent, ≥99.5%
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
Citric acid, 99%
Sigma-Aldrich
Dextrose, meets EP, BP, JP, USP testing specifications, anhydrous
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
D-(+)-Glucose, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
Citric acid, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.5%
Sigma-Aldrich
Citric acid, anhydrous, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
Citric acid, BioUltra, anhydrous, ≥99.5% (T)
Sigma-Aldrich
Citric acid, ≥99.5%, FCC, FG
Sigma-Aldrich
2-Hydroxybutyric acid sodium salt, 97%
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C