  • Search Results
  • Conversion of mouse fibroblasts into cardiomyocyte-like cells using small molecule treatments.
Conversion of mouse fibroblasts into cardiomyocyte-like cells using small molecule treatments.
Biomaterials (2015-04-25)
Gyuman Park, Byung Sun Yoon, Yoon Sik Kim, Seung-Cheol Choi, Jai-Hee Moon, Suhyun Kwon, Jihye Hwang, Wonjin Yun, Jong-Ho Kim, Chi-Yeon Park, Do-Sun Lim, Yang In Kim, Chil Hwan Oh, Seungkwon You
ABSTRACT
The possibility of controlling cell fates by overexpressing specific transcription factors has led to numerous studies in stem cell research. Small molecules can be used, instead of transcription factors, to induce the de-differentiation of somatic cells or to induce pluripotent cells (iPSCs). Here we reported that combinations of small molecules could convert mouse fibroblasts into cardiomyocyte-like cell without requiring transcription factor expression. Treatment with specific combinations of small molecules that are enhancer for iPSC induction converted mouse fibroblasts into spontaneously contracting, cardiac troponin T-positive, cardiomyocyte-like cells. We specifically identified five small molecules that can induce mouse fibroblasts to form these cardiomyocyte-like cells. These cells are similar to primary cardiomyocytes in terms of marker gene expression, epigenetic status of cardiac-specific genes, and subcellular structure. Our findings indicate that lineage conversion can be induced not only by transcription factors, but also by small molecules.
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
L-Ascorbic acid, suitable for plant cell culture
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
Sodium orthovanadate, 99.98% trace metals basis
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
Sodium orthovanadate, ≥90% (titration)
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
Sodium pyrophosphate tetrabasic, ≥95%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90%
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
L-Glutamine
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source