• Início
  • Resultados da busca
  • Active Drug Targeting of a Folate-Based Cyclodextrin-Doxorubicin Conjugate and the Cytotoxic Effect on Drug-Resistant Mammary Tumor Cells In Vitro.
Active Drug Targeting of a Folate-Based Cyclodextrin-Doxorubicin Conjugate and the Cytotoxic Effect on Drug-Resistant Mammary Tumor Cells In Vitro.
Journal of pharmaceutical sciences (2015-05-06)
Hideki Mizusako, Tatsuaki Tagami, Kenjiro Hattori, Tetsuya Ozeki
RESUMO
Active drug targeting is an effective therapeutic approach for the treatment of malignant cancers and novel types of drug carriers have been developed. In this study, we developed a cyclodextrin (CD)-based novel carrier-drug conjugate, called per-FOL-β-CD-ss-DOX, which has folic acid (FA) molecules at the end of primary hydroxyl groups of β-CD and a pH-cleavable spacer with an anticancer drug, doxorubicin (DOX), at the end of secondary hydroxyl groups. This per-FOL-β-CD-ss-DOX exhibited a significant cellular uptake as compared with the free DOX solution by EMT6/P cells, which activate the expression of folate receptor (FR). Cellular uptake of per-FOL-β-CD-ss-DOX was significantly inhibited in the presence of FA and was also inhibited at 4°C. The conjugate exhibited remarkable cytotoxic effects in EMT6/AR1 cells, which are resistant to DOX, whereas free DOX solution did not exhibit this effect. These results suggest that per-FOL-β-CD-ss-DOX can be taken up into cells via FR-related endocytosis and the cleaved DOX derived from it in endosomes could escape the efflux caused by P-glycoprotein, resulting in the cytotoxic effect. Therefore, the drug delivery by per-FOL-β-CD-ss-DOX may be a useful approach for drug delivery to FR-expressing cells such as drug-resistant malignant cancers.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Triethylamine, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Triethylamine, ≥99%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Doxorubicin hydrochloride, 98.0-102.0% (HPLC)
Sigma-Aldrich
Triethylamine, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Folic acid, ≥97%
Sigma-Aldrich
1,4-Diazabicyclo[2.2.2]octane, ReagentPlus®, ≥99%
Sigma-Aldrich
Triethylamine, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Succinic anhydride, ≥99% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Folic acid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥97%
Sigma-Aldrich
Doxorubicin hydrochloride, suitable for fluorescence, 98.0-102.0% (HPLC)
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Dabco® 33-LV
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Folic acid, meets USP testing specifications
Sigma-Aldrich
Triethylamine, for amino acid analysis, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, for protein sequence analysis, ampule, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®